Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 13580744-13580985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TASP1
Entrez Gene ID
55617
Gene Name
taspase 1
Gene Aliases
C20orf13, dJ585I14.2
UniGene
Hs.348297
Gene Chromosome Location
Chr.20: 13104760-13638936 on Build GRCh38
Transcripts
NM_001323602.1 NM_001323603.1 NM_001323604.1 NM_017714.2 NR_136628.1 NR_136629.1 NR_136630.1 NR_136631.1 XM_011529292.1 XM_011529294.1 XM_017027927.1 XM_017027928.1 XM_017027929.1 XM_017027930.1 XM_017027931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5943049 c.404-4_404-3insT p.? Chr.20: 13580984-13580985 on Build GRCh38
5724369 c.481C>T p.P161S Chr.20: 13580904-13580904 on Build GRCh38
5943061 c.404-4_404-3insTT p.? Chr.20: 13580984-13580985 on Build GRCh38
6243392 c.404-4A>T p.? Chr.20: 13580985-13580985 on Build GRCh38
4732239 c.436C>T p.L146F Chr.20: 13580949-13580949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769407513 Chr.20: 13580870-13580870 on Build GRCh38
rs757218445 Chr.20: 13580935-13580935 on Build GRCh38
rs546979957 Chr.20: 13580874-13580874 on Build GRCh38
rs756675887 Chr.20: 13580917-13580917 on Build GRCh38
rs750269503 Chr.20: 13580919-13580919 on Build GRCh38
rs749032971 Chr.20: 13580879-13580879 on Build GRCh38
rs185677724 Chr.20: 13580834-13580834 on Build GRCh38
rs779990596 Chr.20: 13580939-13580939 on Build GRCh38
rs772397933 Chr.20: 13580943-13580943 on Build GRCh38
rs780006701 Chr.20: 13580871-13580871 on Build GRCh38
rs950008422 Chr.20: 13580804-13580804 on Build GRCh38
rs780787096 Chr.20: 13580918-13580918 on Build GRCh38
rs953689833 Chr.20: 13580886-13580886 on Build GRCh38
rs760096126 Chr.20: 13580890-13580890 on Build GRCh38
rs756132529 Chr.20: 13580920-13580920 on Build GRCh38
rs748367651 Chr.20: 13580872-13580872 on Build GRCh38
rs113913531 Chr.20: 13580848-13580848 on Build GRCh38
rs745841812 Chr.20: 13580860-13580860 on Build GRCh38
rs751048210 Chr.20: 13580910-13580910 on Build GRCh38
rs199501456 Chr.20: 13580954-13580954 on Build GRCh38
rs749135048 Chr.20: 13580942-13580942 on Build GRCh38
rs762593097 Chr.20: 13580902-13580902 on Build GRCh38
rs751138224 Chr.20: 13580849-13580849 on Build GRCh38
rs757190546 Chr.20: 13580859-13580860 on Build GRCh38
rs931223829 Chr.20: 13580800-13580800 on Build GRCh38
rs201434844 Chr.20: 13580882-13580882 on Build GRCh38
rs765464426 Chr.20: 13580899-13580899 on Build GRCh38
rs915752212 Chr.20: 13580799-13580799 on Build GRCh38
rs577923955 Chr.20: 13580857-13580857 on Build GRCh38
rs927495671 Chr.20: 13580838-13580838 on Build GRCh38
rs146615455 Chr.20: 13580798-13580798 on Build GRCh38
rs778088119 Chr.20: 13580951-13580951 on Build GRCh38
rs535638432 Chr.20: 13580962-13580962 on Build GRCh38
rs6033761 Chr.20: 13580855-13580855 on Build GRCh38
rs760881777 Chr.20: 13580876-13580876 on Build GRCh38
rs374170761 Chr.20: 13580923-13580923 on Build GRCh38
rs996608944 Chr.20: 13580868-13580868 on Build GRCh38
rs758062443 Chr.20: 13580846-13580846 on Build GRCh38

View Full Product Details