Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 99476336-99476605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF789
Entrez Gene ID
285989
Gene Name
zinc finger protein 789
Gene Aliases
-
UniGene
Hs.440384
Gene Chromosome Location
Chr.7: 99472819-99487600 on Build GRCh38
Transcripts
NM_001013258.1 NM_213603.2 XM_011516067.2 XM_011516070.2 XM_017012018.1 XM_017012019.1 XM_017012020.1 XM_017012021.1 XM_017012023.1 XM_017012024.1 XM_017012025.1 XM_017012026.1 XM_017012027.1

SNP Detail

SNP ID Chromosome Location
rs373312717 Chr.7: 99476497-99476497 on Build GRCh38
rs766247278 Chr.7: 99476421-99476421 on Build GRCh38
rs368930673 Chr.7: 99476516-99476516 on Build GRCh38
rs141232056 Chr.7: 99476377-99476377 on Build GRCh38
rs759367546 Chr.7: 99476512-99476512 on Build GRCh38
rs369685171 Chr.7: 99476487-99476487 on Build GRCh38
rs779259615 Chr.7: 99476437-99476437 on Build GRCh38
rs762508143 Chr.7: 99476414-99476414 on Build GRCh38
rs1039303634 Chr.7: 99476428-99476428 on Build GRCh38
rs767236457 Chr.7: 99476430-99476430 on Build GRCh38
rs750513791 Chr.7: 99476438-99476438 on Build GRCh38
rs962875360 Chr.7: 99476535-99476535 on Build GRCh38
rs549446964 Chr.7: 99476563-99476563 on Build GRCh38
rs747545824 Chr.7: 99476451-99476451 on Build GRCh38
rs748613686 Chr.7: 99476493-99476493 on Build GRCh38
rs372791473 Chr.7: 99476413-99476413 on Build GRCh38
rs1004864972 Chr.7: 99476533-99476535 on Build GRCh38
rs758562045 Chr.7: 99476447-99476447 on Build GRCh38
rs751299739 Chr.7: 99476378-99476378 on Build GRCh38
rs751502188 Chr.7: 99476425-99476425 on Build GRCh38
rs371896349 Chr.7: 99476435-99476435 on Build GRCh38
rs376552016 Chr.7: 99476476-99476476 on Build GRCh38
rs529108710 Chr.7: 99476371-99476371 on Build GRCh38
rs892307461 Chr.7: 99476358-99476358 on Build GRCh38
rs890386799 Chr.7: 99476397-99476397 on Build GRCh38
rs752427899 Chr.7: 99476532-99476532 on Build GRCh38
rs759286403 Chr.7: 99476427-99476427 on Build GRCh38
rs1027432463 Chr.7: 99476579-99476579 on Build GRCh38
rs769527773 Chr.7: 99476408-99476408 on Build GRCh38
rs769257804 Chr.7: 99476481-99476481 on Build GRCh38
rs375032049 Chr.7: 99476506-99476506 on Build GRCh38
rs112029658 Chr.7: 99476498-99476498 on Build GRCh38
rs879145053 Chr.7: 99476503-99476503 on Build GRCh38
rs780669809 Chr.7: 99476448-99476448 on Build GRCh38
rs1015822064 Chr.7: 99476534-99476534 on Build GRCh38
rs959922082 Chr.7: 99476561-99476561 on Build GRCh38

View Full Product Details