Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154325252-154325513 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TKTL1
Entrez Gene ID
8277
Gene Name
transketolase like 1
Gene Aliases
TKR, TKT2
UniGene
Hs.102866
Gene Chromosome Location
Chr.X: 154295674-154330363 on Build GRCh38
Transcripts
NM_001145933.1 NM_001145934.1 NM_012253.3

SNP Detail

SNP ID Chromosome Location
rs782037203 Chr.X: 154325437-154325437 on Build GRCh38
rs186278563 Chr.X: 154325297-154325297 on Build GRCh38
rs781858115 Chr.X: 154325395-154325395 on Build GRCh38
rs782773653 Chr.X: 154325447-154325447 on Build GRCh38
rs368347438 Chr.X: 154325306-154325306 on Build GRCh38
rs782396928 Chr.X: 154325334-154325334 on Build GRCh38
rs974713221 Chr.X: 154325280-154325280 on Build GRCh38
rs4898475 Chr.X: 154325435-154325435 on Build GRCh38
rs782250656 Chr.X: 154325331-154325331 on Build GRCh38
rs936805186 Chr.X: 154325342-154325342 on Build GRCh38
rs1026622387 Chr.X: 154325441-154325441 on Build GRCh38
rs781884803 Chr.X: 154325453-154325453 on Build GRCh38
rs781840319 Chr.X: 154325290-154325290 on Build GRCh38
rs782667299 Chr.X: 154325400-154325400 on Build GRCh38
rs782621324 Chr.X: 154325399-154325399 on Build GRCh38
rs140507131 Chr.X: 154325372-154325372 on Build GRCh38
rs183407036 Chr.X: 154325428-154325428 on Build GRCh38
rs781808942 Chr.X: 154325465-154325465 on Build GRCh38
rs782405084 Chr.X: 154325436-154325436 on Build GRCh38
rs781864611 Chr.X: 154325339-154325339 on Build GRCh38
rs782445587 Chr.X: 154325418-154325418 on Build GRCh38
rs923309587 Chr.X: 154325284-154325284 on Build GRCh38
rs964790513 Chr.X: 154325272-154325272 on Build GRCh38
rs1006449291 Chr.X: 154325452-154325452 on Build GRCh38
rs782433499 Chr.X: 154325311-154325311 on Build GRCh38
rs964757748 Chr.X: 154325462-154325462 on Build GRCh38
rs782116877 Chr.X: 154325445-154325445 on Build GRCh38
rs782162062 Chr.X: 154325454-154325454 on Build GRCh38
rs782223498 Chr.X: 154325427-154325427 on Build GRCh38
rs782796011 Chr.X: 154325301-154325301 on Build GRCh38
rs782197097 Chr.X: 154325426-154325426 on Build GRCh38
rs190698074 Chr.X: 154325341-154325341 on Build GRCh38
rs782339219 Chr.X: 154325439-154325439 on Build GRCh38
rs782581175 Chr.X: 154325315-154325315 on Build GRCh38
rs782530625 Chr.X: 154325297-154325297 on Build GRCh38
rs782108113 Chr.X: 154325360-154325360 on Build GRCh38
rs370025375 Chr.X: 154325410-154325410 on Build GRCh38
rs372785538 Chr.X: 154325444-154325444 on Build GRCh38
rs375077916 Chr.X: 154325457-154325457 on Build GRCh38
rs781801003 Chr.X: 154325375-154325375 on Build GRCh38
rs782764473 Chr.X: 154325362-154325362 on Build GRCh38
rs782618640 Chr.X: 154325325-154325325 on Build GRCh38
rs150438789 Chr.X: 154325401-154325401 on Build GRCh38
rs782464950 Chr.X: 154325386-154325386 on Build GRCh38
rs781964116 Chr.X: 154325351-154325351 on Build GRCh38

View Full Product Details