Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53576817-53577060 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF331
Entrez Gene ID
55422
Gene Name
zinc finger protein 331
Gene Aliases
RITA, ZNF361, ZNF463
UniGene
Hs.185674
Gene Chromosome Location
Chr.19: 53503819-53580269 on Build GRCh38
Transcripts
NM_001079906.1 NM_001079907.1 NM_001253798.1 NM_001253799.1 NM_001253800.1 NM_001253801.2 NM_001317113.1 NM_001317114.1 NM_001317115.1 NM_001317116.1 NM_001317117.1 NM_001317118.1 NM_001317119.1 NM_001317120.1 NM_001317121.1 NM_018555.5 XM_011527076.2 XM_011527078.2 XM_017026936.1 XM_017026937.1 XM_017026938.1 XM_017026939.1 XM_017026940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5515737 c.324C>A p.V108V Chr.19: 53576884-53576884 on Build GRCh38
3893327 c.381G>A p.K127K Chr.19: 53576941-53576941 on Build GRCh38
6829392 c.295T>G p.Y99D Chr.19: 53576855-53576855 on Build GRCh38
475221 c.447T>G p.H149Q Chr.19: 53577007-53577007 on Build GRCh38
3783286 c.317A>T p.N106I Chr.19: 53576877-53576877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs8109631 Chr.19: 53576890-53576890 on Build GRCh38
rs749796126 Chr.19: 53576911-53576911 on Build GRCh38
rs968408298 Chr.19: 53577028-53577028 on Build GRCh38
rs369494593 Chr.19: 53576841-53576841 on Build GRCh38
rs772690900 Chr.19: 53576922-53576922 on Build GRCh38
rs747179966 Chr.19: 53576866-53576866 on Build GRCh38
rs954574618 Chr.19: 53576896-53576896 on Build GRCh38
rs765129221 Chr.19: 53576960-53576960 on Build GRCh38
rs766379010 Chr.19: 53576939-53576941 on Build GRCh38
rs760274215 Chr.19: 53576924-53576924 on Build GRCh38
rs368241758 Chr.19: 53576913-53576913 on Build GRCh38
rs764174432 Chr.19: 53576994-53576994 on Build GRCh38
rs112855712 Chr.19: 53576886-53576886 on Build GRCh38
rs752642053 Chr.19: 53576969-53576969 on Build GRCh38
rs775913323 Chr.19: 53576898-53576898 on Build GRCh38
rs758433153 Chr.19: 53576985-53576985 on Build GRCh38
rs751560573 Chr.19: 53577015-53577015 on Build GRCh38
rs758486389 Chr.19: 53576840-53576840 on Build GRCh38
rs1053617488 Chr.19: 53576919-53576919 on Build GRCh38
rs776403470 Chr.19: 53576937-53576937 on Build GRCh38
rs757396686 Chr.19: 53576877-53576877 on Build GRCh38
rs1351 Chr.19: 53577025-53577025 on Build GRCh38
rs34867717 Chr.19: 53576953-53576954 on Build GRCh38
rs150965267 Chr.19: 53577016-53577016 on Build GRCh38
rs765946542 Chr.19: 53576925-53576925 on Build GRCh38
rs372185704 Chr.19: 53576873-53576873 on Build GRCh38

View Full Product Details