Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2933945-2934190 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF77
Entrez Gene ID
58492
Gene Name
zinc finger protein 77
Gene Aliases
pT1
UniGene
Hs.38004
Gene Chromosome Location
Chr.19: 2933218-2944971 on Build GRCh38
Transcripts
NM_021217.2 XM_017027081.1 XM_017027082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1392311 c.1095C>A p.S365S Chr.19: 2934032-2934032 on Build GRCh38
3742878 c.951G>A p.T317T Chr.19: 2934176-2934176 on Build GRCh38
7338021 c.1174C>T p.P392S Chr.19: 2933953-2933953 on Build GRCh38
4888671 c.1168G>T p.G390* Chr.19: 2933959-2933959 on Build GRCh38
1271770 c.1113G>A p.M371I Chr.19: 2934014-2934014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771819659 Chr.19: 2933988-2933988 on Build GRCh38
rs779911205 Chr.19: 2933992-2933992 on Build GRCh38
rs756894144 Chr.19: 2934035-2934035 on Build GRCh38
rs574952125 Chr.19: 2934116-2934116 on Build GRCh38
rs142377561 Chr.19: 2934075-2934075 on Build GRCh38
rs199562140 Chr.19: 2934093-2934093 on Build GRCh38
rs754838425 Chr.19: 2934137-2934137 on Build GRCh38
rs868523598 Chr.19: 2934117-2934117 on Build GRCh38
rs766389851 Chr.19: 2934131-2934131 on Build GRCh38
rs375886688 Chr.19: 2934055-2934055 on Build GRCh38
rs749978906 Chr.19: 2934123-2934123 on Build GRCh38
rs866343412 Chr.19: 2934099-2934099 on Build GRCh38
rs200559103 Chr.19: 2934144-2934144 on Build GRCh38
rs370214322 Chr.19: 2934058-2934058 on Build GRCh38
rs879131348 Chr.19: 2933987-2933987 on Build GRCh38
rs372777520 Chr.19: 2934092-2934092 on Build GRCh38
rs763884242 Chr.19: 2934030-2934030 on Build GRCh38
rs745654235 Chr.19: 2933978-2933978 on Build GRCh38
rs776644244 Chr.19: 2934007-2934007 on Build GRCh38
rs17854297 Chr.19: 2934056-2934056 on Build GRCh38
rs773779717 Chr.19: 2934018-2934018 on Build GRCh38
rs759981598 Chr.19: 2934102-2934102 on Build GRCh38
rs1021914057 Chr.19: 2934041-2934041 on Build GRCh38
rs201631747 Chr.19: 2934052-2934052 on Build GRCh38
rs200311160 Chr.19: 2934132-2934132 on Build GRCh38
rs781383879 Chr.19: 2934138-2934138 on Build GRCh38
rs868580731 Chr.19: 2934133-2934133 on Build GRCh38
rs535918404 Chr.19: 2934015-2934015 on Build GRCh38
rs368804533 Chr.19: 2934082-2934082 on Build GRCh38
rs768739092 Chr.19: 2934066-2934066 on Build GRCh38
rs763365592 Chr.19: 2934022-2934022 on Build GRCh38
rs560203335 Chr.19: 2934163-2934163 on Build GRCh38
rs138948804 Chr.19: 2934140-2934140 on Build GRCh38
rs61740279 Chr.19: 2934025-2934025 on Build GRCh38
rs374136791 Chr.19: 2934072-2934072 on Build GRCh38
rs772507748 Chr.19: 2934165-2934165 on Build GRCh38
rs943443756 Chr.19: 2933974-2933974 on Build GRCh38
rs774985424 Chr.19: 2934160-2934160 on Build GRCh38
rs749694668 Chr.19: 2934083-2934083 on Build GRCh38
rs372280417 Chr.19: 2934127-2934127 on Build GRCh38
rs374049271 Chr.19: 2934024-2934024 on Build GRCh38
rs866456026 Chr.19: 2934112-2934112 on Build GRCh38
rs757235143 Chr.19: 2933975-2933975 on Build GRCh38
rs1009463811 Chr.19: 2934036-2934036 on Build GRCh38
rs747132521 Chr.19: 2934001-2934001 on Build GRCh38
rs779030199 Chr.19: 2933977-2933977 on Build GRCh38
rs768955579 Chr.19: 2934002-2934002 on Build GRCh38
rs763830961 Chr.19: 2934108-2934108 on Build GRCh38
rs146879198 Chr.19: 2934109-2934109 on Build GRCh38
rs761931172 Chr.19: 2934009-2934009 on Build GRCh38
rs781433172 Chr.19: 2934067-2934067 on Build GRCh38
rs199748725 Chr.19: 2934031-2934031 on Build GRCh38
rs753363434 Chr.19: 2934033-2934033 on Build GRCh38
rs903553128 Chr.19: 2934042-2934042 on Build GRCh38
rs369325243 Chr.19: 2934151-2934151 on Build GRCh38
rs764771353 Chr.19: 2934122-2934122 on Build GRCh38
rs756221080 Chr.19: 2934015-2934016 on Build GRCh38
rs149394326 Chr.19: 2934149-2934149 on Build GRCh38
rs370544700 Chr.19: 2934017-2934017 on Build GRCh38
rs779674823 Chr.19: 2934057-2934057 on Build GRCh38
rs753418407 Chr.19: 2933971-2933971 on Build GRCh38
rs542089630 Chr.19: 2934152-2934152 on Build GRCh38

View Full Product Details