Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 63727721-63727950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SGPP1
Entrez Gene ID
81537
Gene Name
sphingosine-1-phosphate phosphatase 1
Gene Aliases
SPPase1
UniGene
Hs.24678
Gene Chromosome Location
Chr.14: 63684216-63728095 on Build GRCh38
Transcripts
NM_030791.3 XM_017021678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4148159 c.155A>C p.D52A Chr.14: 63727790-63727790 on Build GRCh38
6380618 c.92C>T p.A31V Chr.14: 63727853-63727853 on Build GRCh38
6434525 c.110C>T p.A37V Chr.14: 63727835-63727835 on Build GRCh38
6270099 c.92C>A p.A31E Chr.14: 63727853-63727853 on Build GRCh38
6428300 c.148G>A p.A50T Chr.14: 63727797-63727797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023743518 Chr.14: 63727834-63727834 on Build GRCh38
rs762638770 Chr.14: 63727828-63727828 on Build GRCh38
rs1031453037 Chr.14: 63727897-63727897 on Build GRCh38
rs752288586 Chr.14: 63727839-63727839 on Build GRCh38
rs755217669 Chr.14: 63727912-63727912 on Build GRCh38
rs1036406295 Chr.14: 63727774-63727774 on Build GRCh38
rs765677196 Chr.14: 63727856-63727856 on Build GRCh38
rs1056711086 Chr.14: 63727822-63727822 on Build GRCh38
rs766263016 Chr.14: 63727829-63727829 on Build GRCh38
rs759309325 Chr.14: 63727833-63727833 on Build GRCh38
rs868082996 Chr.14: 63727929-63727929 on Build GRCh38
rs897483492 Chr.14: 63727787-63727787 on Build GRCh38
rs751762148 Chr.14: 63727908-63727908 on Build GRCh38
rs772998098 Chr.14: 63727789-63727789 on Build GRCh38
rs529267096 Chr.14: 63727747-63727747 on Build GRCh38
rs750996925 Chr.14: 63727864-63727864 on Build GRCh38
rs531814617 Chr.14: 63727881-63727881 on Build GRCh38
rs865973433 Chr.14: 63727910-63727910 on Build GRCh38
rs200531351 Chr.14: 63727835-63727835 on Build GRCh38
rs757772840 Chr.14: 63727849-63727849 on Build GRCh38
rs149126697 Chr.14: 63727811-63727811 on Build GRCh38
rs8006901 Chr.14: 63727741-63727741 on Build GRCh38
rs773843910 Chr.14: 63727830-63727830 on Build GRCh38
rs780511156 Chr.14: 63727891-63727891 on Build GRCh38
rs769724041 Chr.14: 63727757-63727757 on Build GRCh38

View Full Product Details