Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186801028-186801289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC4
Entrez Gene ID
5984
Gene Name
replication factor C subunit 4
Gene Aliases
A1, RFC37
UniGene
Hs.732098
Gene Chromosome Location
Chr.3: 186789892-186806695 on Build GRCh38
Transcripts
NM_002916.3 NM_181573.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
446161 c.203G>C p.G68A Chr.3: 186801124-186801124 on Build GRCh38
4721146 c.192_193insA p.S65fs*9 Chr.3: 186801134-186801135 on Build GRCh38
5113489 c.132-6C>T p.? Chr.3: 186801201-186801201 on Build GRCh38
3723347 c.189A>C p.K63N Chr.3: 186801138-186801138 on Build GRCh38
6868670 c.162C>T p.F54F Chr.3: 186801165-186801165 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs753783647 Chr.3: 186801149-186801149 on Build GRCh38
rs151335809 Chr.3: 186801146-186801146 on Build GRCh38
rs776497631 Chr.3: 186801230-186801230 on Build GRCh38
rs918767772 Chr.3: 186801245-186801245 on Build GRCh38
rs750354323 Chr.3: 186801234-186801234 on Build GRCh38
rs765251058 Chr.3: 186801233-186801233 on Build GRCh38
rs776828891 Chr.3: 186801134-186801135 on Build GRCh38
rs775176913 Chr.3: 186801222-186801222 on Build GRCh38
rs140654716 Chr.3: 186801152-186801152 on Build GRCh38
rs763514547 Chr.3: 186801067-186801067 on Build GRCh38
rs973363522 Chr.3: 186801236-186801236 on Build GRCh38
rs749560125 Chr.3: 186801201-186801201 on Build GRCh38
rs371705418 Chr.3: 186801223-186801223 on Build GRCh38
rs751680916 Chr.3: 186801170-186801170 on Build GRCh38
rs767192098 Chr.3: 186801080-186801080 on Build GRCh38
rs773928424 Chr.3: 186801075-186801075 on Build GRCh38
rs1046094886 Chr.3: 186801161-186801161 on Build GRCh38
rs770351072 Chr.3: 186801199-186801199 on Build GRCh38
rs752760679 Chr.3: 186801240-186801240 on Build GRCh38
rs181497505 Chr.3: 186801054-186801054 on Build GRCh38
rs755167691 Chr.3: 186801239-186801239 on Build GRCh38
rs771493245 Chr.3: 186801204-186801204 on Build GRCh38
rs185005078 Chr.3: 186801231-186801231 on Build GRCh38
rs755069325 Chr.3: 186801178-186801178 on Build GRCh38
rs765551839 Chr.3: 186801102-186801103 on Build GRCh38
rs778205938 Chr.3: 186801200-186801200 on Build GRCh38
rs759000132 Chr.3: 186801078-186801078 on Build GRCh38
rs774685220 Chr.3: 186801206-186801206 on Build GRCh38
rs144428719 Chr.3: 186801169-186801169 on Build GRCh38
rs762284946 Chr.3: 186801235-186801235 on Build GRCh38
rs752318851 Chr.3: 186801118-186801118 on Build GRCh38
rs781615742 Chr.3: 186801184-186801184 on Build GRCh38
rs201686899 Chr.3: 186801122-186801122 on Build GRCh38
rs199657013 Chr.3: 186801243-186801244 on Build GRCh38
rs763583798 Chr.3: 186801225-186801225 on Build GRCh38
rs748367842 Chr.3: 186801198-186801198 on Build GRCh38
rs771545124 Chr.3: 186801215-186801215 on Build GRCh38
rs771277725 Chr.3: 186801072-186801072 on Build GRCh38

View Full Product Details