Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35032313-35032584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCN1B
Entrez Gene ID
6324
Gene Name
sodium voltage-gated channel beta subunit 1
Gene Aliases
ATFB13, BRGDA5, GEFSP1
UniGene
Hs.436646
Gene Chromosome Location
Chr.19: 35030688-35040449 on Build GRCh38
Transcripts
NM_001037.4 NM_001321605.1 NM_199037.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3422668 c.77C>T p.S26L Chr.19: 35032564-35032564 on Build GRCh38
5592991 c.90C>T p.A30A Chr.19: 35032577-35032577 on Build GRCh38
6899461 c.57G>A p.G19G Chr.19: 35032544-35032544 on Build GRCh38
3532420 c.59G>A p.G20D Chr.19: 35032546-35032546 on Build GRCh38
5592992 c.90C>T p.A30A Chr.19: 35032577-35032577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758499324 Chr.19: 35032386-35032386 on Build GRCh38
rs745578106 Chr.19: 35032551-35032551 on Build GRCh38
rs762553865 Chr.19: 35032556-35032556 on Build GRCh38
rs535122676 Chr.19: 35032462-35032463 on Build GRCh38
rs768653839 Chr.19: 35032564-35032564 on Build GRCh38
rs769410262 Chr.19: 35032553-35032553 on Build GRCh38
rs754980960 Chr.19: 35032498-35032498 on Build GRCh38
rs766495205 Chr.19: 35032482-35032482 on Build GRCh38
rs749373268 Chr.19: 35032501-35032506 on Build GRCh38
rs140219033 Chr.19: 35032448-35032448 on Build GRCh38
rs1053593283 Chr.19: 35032387-35032387 on Build GRCh38
rs746930325 Chr.19: 35032353-35032353 on Build GRCh38
rs376338621 Chr.19: 35032488-35032488 on Build GRCh38
rs138759136 Chr.19: 35032364-35032364 on Build GRCh38
rs562094912 Chr.19: 35032439-35032439 on Build GRCh38
rs774401364 Chr.19: 35032565-35032565 on Build GRCh38
rs914825430 Chr.19: 35032381-35032381 on Build GRCh38
rs1005312276 Chr.19: 35032541-35032541 on Build GRCh38
rs780469360 Chr.19: 35032487-35032497 on Build GRCh38
rs761678396 Chr.19: 35032480-35032480 on Build GRCh38
rs1044568918 Chr.19: 35032438-35032438 on Build GRCh38
rs200339565 Chr.19: 35032550-35032550 on Build GRCh38
rs374254905 Chr.19: 35032507-35032507 on Build GRCh38
rs970395261 Chr.19: 35032338-35032339 on Build GRCh38
rs905948607 Chr.19: 35032456-35032456 on Build GRCh38
rs752796919 Chr.19: 35032505-35032505 on Build GRCh38
rs370882059 Chr.19: 35032490-35032490 on Build GRCh38
rs786205837 Chr.19: 35032560-35032560 on Build GRCh38
rs531824397 Chr.19: 35032375-35032375 on Build GRCh38
rs779012549 Chr.19: 35032502-35032502 on Build GRCh38
rs778258172 Chr.19: 35032514-35032514 on Build GRCh38
rs773216135 Chr.19: 35032484-35032484 on Build GRCh38
rs771386831 Chr.19: 35032542-35032542 on Build GRCh38
rs548443747 Chr.19: 35032417-35032417 on Build GRCh38
rs747279999 Chr.19: 35032523-35032523 on Build GRCh38
rs932730921 Chr.19: 35032443-35032443 on Build GRCh38
rs777983530 Chr.19: 35032424-35032424 on Build GRCh38
rs775096415 Chr.19: 35032554-35032554 on Build GRCh38

View Full Product Details