Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 90270147-90270404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSMC1
Entrez Gene ID
5700
Gene Name
proteasome 26S subunit, ATPase 1
Gene Aliases
P26S4, S4, p56
UniGene
Hs.356654
Gene Chromosome Location
Chr.14: 90256550-90272625 on Build GRCh38
Transcripts
NM_002802.2 XM_017021470.1 XM_017021471.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5546991 c.1059delC p.L355fs*18 Chr.14: 90270223-90270223 on Build GRCh38
3401510 c.1102C>T p.H368Y Chr.14: 90270266-90270266 on Build GRCh38
4052882 c.1083G>C p.K361N Chr.14: 90270247-90270247 on Build GRCh38
363263 c.1037G>A p.R346H Chr.14: 90270201-90270201 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933216440 Chr.14: 90270220-90270220 on Build GRCh38
rs781271442 Chr.14: 90270244-90270244 on Build GRCh38
rs888987761 Chr.14: 90270257-90270257 on Build GRCh38
rs753090098 Chr.14: 90270204-90270204 on Build GRCh38
rs759012045 Chr.14: 90270216-90270216 on Build GRCh38
rs770542358 Chr.14: 90270295-90270295 on Build GRCh38
rs907691551 Chr.14: 90270240-90270240 on Build GRCh38
rs532770063 Chr.14: 90270235-90270235 on Build GRCh38
rs765756207 Chr.14: 90270192-90270192 on Build GRCh38
rs772023510 Chr.14: 90270282-90270282 on Build GRCh38
rs778723948 Chr.14: 90270268-90270268 on Build GRCh38
rs752034051 Chr.14: 90270367-90270367 on Build GRCh38
rs764521026 Chr.14: 90270366-90270366 on Build GRCh38
rs756313118 Chr.14: 90270256-90270256 on Build GRCh38
rs759258081 Chr.14: 90270169-90270169 on Build GRCh38
rs746153150 Chr.14: 90270248-90270248 on Build GRCh38
rs987986953 Chr.14: 90270270-90270270 on Build GRCh38
rs922635500 Chr.14: 90270194-90270194 on Build GRCh38
rs776027100 Chr.14: 90270347-90270347 on Build GRCh38
rs746507139 Chr.14: 90270285-90270285 on Build GRCh38
rs757300947 Chr.14: 90270233-90270233 on Build GRCh38
rs566274159 Chr.14: 90270337-90270337 on Build GRCh38
rs940364305 Chr.14: 90270179-90270179 on Build GRCh38
rs374587393 Chr.14: 90270184-90270184 on Build GRCh38
rs1009225987 Chr.14: 90270284-90270284 on Build GRCh38
rs759313248 Chr.14: 90270320-90270320 on Build GRCh38
rs898937548 Chr.14: 90270191-90270191 on Build GRCh38
rs1036036719 Chr.14: 90270188-90270188 on Build GRCh38
rs757695727 Chr.14: 90270370-90270370 on Build GRCh38
rs751612223 Chr.14: 90270228-90270228 on Build GRCh38
rs764620445 Chr.14: 90270221-90270221 on Build GRCh38
rs150699564 Chr.14: 90270307-90270307 on Build GRCh38
rs747900278 Chr.14: 90270271-90270271 on Build GRCh38
rs183723361 Chr.14: 90270276-90270276 on Build GRCh38
rs367830385 Chr.14: 90270283-90270283 on Build GRCh38
rs763360590 Chr.14: 90270365-90270365 on Build GRCh38

View Full Product Details