Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1234030-1234273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD21L1
Entrez Gene ID
642636
Gene Name
RAD21 cohesin complex component like 1
Gene Aliases
RAD21L, dJ545L17.2
UniGene
Hs.713451
Gene Chromosome Location
Chr.20: 1226043-1255876 on Build GRCh38
Transcripts
NM_001136566.2 XM_006723602.2 XM_006723603.2 XM_006723604.2 XM_006723605.3 XM_011529317.2 XM_011529319.1 XM_011529320.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6372626 c.390C>G p.H130Q Chr.20: 1234106-1234106 on Build GRCh38
6208408 c.475+1G>A p.? Chr.20: 1234192-1234192 on Build GRCh38
6357675 c.454A>G p.I152V Chr.20: 1234170-1234170 on Build GRCh38
6208407 c.475+1G>A p.? Chr.20: 1234192-1234192 on Build GRCh38
6372625 c.390C>G p.H130Q Chr.20: 1234106-1234106 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752838571 Chr.20: 1234109-1234109 on Build GRCh38
rs552283610 Chr.20: 1234151-1234151 on Build GRCh38
rs954879369 Chr.20: 1234101-1234101 on Build GRCh38
rs755285899 Chr.20: 1234085-1234085 on Build GRCh38
rs373813678 Chr.20: 1234238-1234238 on Build GRCh38
rs537732457 Chr.20: 1234221-1234221 on Build GRCh38
rs750283489 Chr.20: 1234092-1234092 on Build GRCh38
rs765509960 Chr.20: 1234089-1234089 on Build GRCh38
rs942425695 Chr.20: 1234219-1234219 on Build GRCh38
rs203534 Chr.20: 1234170-1234170 on Build GRCh38
rs1022045414 Chr.20: 1234065-1234065 on Build GRCh38
rs566589485 Chr.20: 1234079-1234079 on Build GRCh38
rs760315180 Chr.20: 1234081-1234081 on Build GRCh38
rs532371215 Chr.20: 1234148-1234148 on Build GRCh38
rs186913953 Chr.20: 1234224-1234224 on Build GRCh38
rs1029261120 Chr.20: 1234083-1234083 on Build GRCh38
rs1016475433 Chr.20: 1234093-1234093 on Build GRCh38
rs749041115 Chr.20: 1234236-1234236 on Build GRCh38
rs755837382 Chr.20: 1234165-1234165 on Build GRCh38

View Full Product Details