Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228148394-228148659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUK1
Entrez Gene ID
2987
Gene Name
guanylate kinase 1
Gene Aliases
GMK
UniGene
Hs.376933
Gene Chromosome Location
Chr.1: 228139643-228148955 on Build GRCh38
Transcripts
NM_000858.5 NM_001159390.1 NM_001159391.1 NM_001242839.1 NM_001242840.1 XM_005273103.3 XM_005273104.4 XM_005273105.3 XM_005273106.2 XM_011544174.2 XM_017001091.1 XM_017001092.1 XM_017001093.1 XM_017001094.1 XM_017001095.1 XM_017001096.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4927218 c.515A>G p.D172G Chr.1: 228148410-228148410 on Build GRCh38
7170006 c.551C>T p.S184F Chr.1: 228148452-228148452 on Build GRCh38
7122011 c.636G>A p.L212L Chr.1: 228148537-228148537 on Build GRCh38
5036265 c.536T>C p.L179P Chr.1: 228148437-228148437 on Build GRCh38
4927219 c.509A>G p.D170G Chr.1: 228148410-228148410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202143318 Chr.1: 228148446-228148446 on Build GRCh38
rs574849703 Chr.1: 228148576-228148576 on Build GRCh38
rs139322242 Chr.1: 228148430-228148430 on Build GRCh38
rs763261614 Chr.1: 228148581-228148581 on Build GRCh38
rs777159704 Chr.1: 228148540-228148540 on Build GRCh38
rs372383082 Chr.1: 228148478-228148478 on Build GRCh38
rs994961074 Chr.1: 228148458-228148458 on Build GRCh38
rs767385637 Chr.1: 228148638-228148638 on Build GRCh38
rs774306171 Chr.1: 228148582-228148582 on Build GRCh38
rs771349393 Chr.1: 228148531-228148531 on Build GRCh38
rs374498638 Chr.1: 228148491-228148491 on Build GRCh38
rs988751046 Chr.1: 228148629-228148629 on Build GRCh38
rs542934700 Chr.1: 228148493-228148493 on Build GRCh38
rs775802390 Chr.1: 228148450-228148450 on Build GRCh38
rs150006242 Chr.1: 228148590-228148590 on Build GRCh38
rs369865694 Chr.1: 228148476-228148476 on Build GRCh38
rs553156978 Chr.1: 228148590-228148590 on Build GRCh38
rs766237635 Chr.1: 228148424-228148424 on Build GRCh38
rs763170849 Chr.1: 228148452-228148452 on Build GRCh38
rs750223120 Chr.1: 228148482-228148482 on Build GRCh38
rs778274946 Chr.1: 228148505-228148505 on Build GRCh38
rs957140990 Chr.1: 228148516-228148516 on Build GRCh38
rs770257101 Chr.1: 228148564-228148564 on Build GRCh38
rs746260549 Chr.1: 228148548-228148548 on Build GRCh38
rs1012846738 Chr.1: 228148460-228148460 on Build GRCh38
rs146725528 Chr.1: 228148429-228148429 on Build GRCh38
rs764048018 Chr.1: 228148461-228148461 on Build GRCh38
rs574202623 Chr.1: 228148490-228148490 on Build GRCh38
rs751578817 Chr.1: 228148474-228148474 on Build GRCh38
rs554066960 Chr.1: 228148447-228148447 on Build GRCh38
rs761884906 Chr.1: 228148615-228148615 on Build GRCh38
rs956679020 Chr.1: 228148607-228148607 on Build GRCh38
rs753021136 Chr.1: 228148423-228148423 on Build GRCh38
rs556462695 Chr.1: 228148522-228148522 on Build GRCh38
rs754584696 Chr.1: 228148495-228148495 on Build GRCh38

View Full Product Details