Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89339131-89339366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT3
Entrez Gene ID
3437
Gene Name
interferon induced protein with tetratricopeptide repeats 3
Gene Aliases
CIG-49, GARG-49, IFI60, IFIT4, IRG2, ISG60, P60, RIG-G, cig41
UniGene
Hs.47338
Gene Chromosome Location
Chr.10: 89327819-89340968 on Build GRCh38
Transcripts
NM_001031683.3 NM_001289758.1 NM_001289759.1 NM_001549.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5738979 c.559C>T p.P187S Chr.10: 89339214-89339214 on Build GRCh38
40187 c.640G>A p.G214S Chr.10: 89339295-89339295 on Build GRCh38
5909011 c.610C>T p.P204S Chr.10: 89339265-89339265 on Build GRCh38
428105 c.699C>A p.A233A Chr.10: 89339354-89339354 on Build GRCh38
1210269 c.516C>A p.F172L Chr.10: 89339171-89339171 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762482947 Chr.10: 89339183-89339183 on Build GRCh38
rs540964847 Chr.10: 89339321-89339321 on Build GRCh38
rs146881667 Chr.10: 89339159-89339159 on Build GRCh38
rs749146942 Chr.10: 89339297-89339297 on Build GRCh38
rs144424377 Chr.10: 89339192-89339192 on Build GRCh38
rs139732815 Chr.10: 89339184-89339184 on Build GRCh38
rs763548045 Chr.10: 89339250-89339251 on Build GRCh38
rs770813145 Chr.10: 89339305-89339305 on Build GRCh38
rs774290262 Chr.10: 89339320-89339320 on Build GRCh38
rs78965025 Chr.10: 89339284-89339284 on Build GRCh38
rs775119341 Chr.10: 89339336-89339336 on Build GRCh38
rs762016100 Chr.10: 89339211-89339211 on Build GRCh38
rs142127477 Chr.10: 89339280-89339280 on Build GRCh38
rs772960365 Chr.10: 89339162-89339162 on Build GRCh38
rs765699153 Chr.10: 89339222-89339222 on Build GRCh38
rs376728166 Chr.10: 89339325-89339325 on Build GRCh38
rs930739215 Chr.10: 89339273-89339273 on Build GRCh38
rs769648343 Chr.10: 89339160-89339160 on Build GRCh38
rs755130984 Chr.10: 89339275-89339275 on Build GRCh38
rs775136197 Chr.10: 89339194-89339194 on Build GRCh38
rs146776701 Chr.10: 89339227-89339227 on Build GRCh38
rs773888170 Chr.10: 89339191-89339191 on Build GRCh38
rs755860041 Chr.10: 89339286-89339286 on Build GRCh38
rs749317140 Chr.10: 89339177-89339177 on Build GRCh38
rs758994651 Chr.10: 89339228-89339228 on Build GRCh38
rs754020242 Chr.10: 89339288-89339288 on Build GRCh38
rs149187349 Chr.10: 89339271-89339271 on Build GRCh38
rs375379157 Chr.10: 89339279-89339279 on Build GRCh38
rs766537327 Chr.10: 89339247-89339247 on Build GRCh38

View Full Product Details