Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79300579-79300818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMY
Entrez Gene ID
133746
Gene Name
junction mediating and regulatory protein, p53 cofactor
Gene Aliases
WHAMM2, WHDC1L3
UniGene
Hs.482605
Gene Chromosome Location
Chr.5: 79236102-79327215 on Build GRCh38
Transcripts
NM_152405.4 XM_005248430.2 XM_011543155.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4531821 c.766G>T p.E256* Chr.5: 79300810-79300810 on Build GRCh38
5960086 c.730G>A p.E244K Chr.5: 79300774-79300774 on Build GRCh38
6394114 c.1822C>A p.Q608K Chr.5: 79300804-79300804 on Build GRCh38
5915409 c.1819C>T p.R607C Chr.5: 79300801-79300801 on Build GRCh38
4655372 c.1785G>T p.Q595H Chr.5: 79300767-79300767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs797011875 Chr.5: 79300738-79300738 on Build GRCh38
rs766427064 Chr.5: 79300750-79300750 on Build GRCh38
rs775383072 Chr.5: 79300663-79300663 on Build GRCh38
rs763530665 Chr.5: 79300681-79300681 on Build GRCh38
rs775178471 Chr.5: 79300670-79300670 on Build GRCh38
rs750619880 Chr.5: 79300719-79300719 on Build GRCh38
rs762180475 Chr.5: 79300700-79300700 on Build GRCh38
rs966589599 Chr.5: 79300777-79300777 on Build GRCh38
rs763776390 Chr.5: 79300764-79300766 on Build GRCh38
rs976210166 Chr.5: 79300720-79300720 on Build GRCh38
rs957784206 Chr.5: 79300740-79300740 on Build GRCh38
rs12109475 Chr.5: 79300757-79300757 on Build GRCh38
rs542140622 Chr.5: 79300797-79300797 on Build GRCh38
rs756333244 Chr.5: 79300726-79300726 on Build GRCh38
rs545917331 Chr.5: 79300624-79300624 on Build GRCh38
rs746936628 Chr.5: 79300758-79300758 on Build GRCh38
rs760423547 Chr.5: 79300753-79300754 on Build GRCh38
rs769305062 Chr.5: 79300652-79300652 on Build GRCh38
rs377043963 Chr.5: 79300689-79300689 on Build GRCh38
rs757030849 Chr.5: 79300765-79300765 on Build GRCh38
rs745526562 Chr.5: 79300636-79300636 on Build GRCh38
rs753972990 Chr.5: 79300756-79300756 on Build GRCh38
rs1018268182 Chr.5: 79300673-79300673 on Build GRCh38
rs12109473 Chr.5: 79300698-79300698 on Build GRCh38
rs764755071 Chr.5: 79300687-79300687 on Build GRCh38
rs527666699 Chr.5: 79300748-79300748 on Build GRCh38
rs767796449 Chr.5: 79300710-79300710 on Build GRCh38
rs780882767 Chr.5: 79300766-79300766 on Build GRCh38
rs745504604 Chr.5: 79300767-79300767 on Build GRCh38

View Full Product Details