Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 32594841-32595090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3M
Entrez Gene ID
10480
Gene Name
eukaryotic translation initiation factor 3 subunit M
Gene Aliases
B5, GA17, PCID1, TANGO7, hfl-B5
UniGene
Hs.502244
Gene Chromosome Location
Chr.11: 32583767-32602873 on Build GRCh38
Transcripts
NM_001307929.1 NM_006360.5 XM_011519840.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
687512 c.629G>T p.R210L Chr.11: 32594925-32594925 on Build GRCh38
3396013 c.661G>T p.D221Y Chr.11: 32594957-32594957 on Build GRCh38
4032392 c.703G>A p.E235K Chr.11: 32594999-32594999 on Build GRCh38
6257161 c.700G>T p.G234C Chr.11: 32594996-32594996 on Build GRCh38
2114625 c.713A>G p.H238R Chr.11: 32595009-32595009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372564627 Chr.11: 32594945-32594945 on Build GRCh38
rs749143881 Chr.11: 32595018-32595018 on Build GRCh38
rs770623852 Chr.11: 32595024-32595024 on Build GRCh38
rs779851517 Chr.11: 32595016-32595021 on Build GRCh38
rs777546451 Chr.11: 32595014-32595014 on Build GRCh38
rs756103511 Chr.11: 32595012-32595012 on Build GRCh38
rs757533838 Chr.11: 32594948-32594948 on Build GRCh38
rs771645441 Chr.11: 32595042-32595042 on Build GRCh38
rs377461075 Chr.11: 32595009-32595009 on Build GRCh38
rs758639052 Chr.11: 32594977-32594977 on Build GRCh38
rs766999907 Chr.11: 32594930-32594930 on Build GRCh38
rs12361697 Chr.11: 32595055-32595055 on Build GRCh38
rs761172968 Chr.11: 32594942-32594942 on Build GRCh38
rs756018168 Chr.11: 32594894-32594895 on Build GRCh38
rs1050047695 Chr.11: 32594925-32594925 on Build GRCh38
rs773840093 Chr.11: 32594904-32594904 on Build GRCh38
rs750661301 Chr.11: 32594952-32594952 on Build GRCh38
rs764701575 Chr.11: 32595048-32595048 on Build GRCh38
rs753370368 Chr.11: 32595028-32595029 on Build GRCh38
rs777001290 Chr.11: 32595050-32595050 on Build GRCh38
rs944282076 Chr.11: 32594983-32594983 on Build GRCh38
rs765562096 Chr.11: 32594949-32594949 on Build GRCh38
rs866120558 Chr.11: 32594969-32594969 on Build GRCh38
rs754280375 Chr.11: 32594947-32594947 on Build GRCh38
rs745532025 Chr.11: 32595032-32595032 on Build GRCh38
rs370641696 Chr.11: 32595047-32595047 on Build GRCh38
rs267602853 Chr.11: 32595000-32595000 on Build GRCh38
rs762297252 Chr.11: 32595057-32595057 on Build GRCh38
rs780289523 Chr.11: 32594999-32594999 on Build GRCh38
rs754659944 Chr.11: 32595032-32595033 on Build GRCh38
rs775011347 Chr.11: 32595045-32595045 on Build GRCh38
rs774047095 Chr.11: 32595027-32595027 on Build GRCh38
rs527437764 Chr.11: 32594938-32594938 on Build GRCh38
rs891345103 Chr.11: 32594933-32594933 on Build GRCh38
rs139505078 Chr.11: 32594923-32594923 on Build GRCh38

View Full Product Details