Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58523281-58523554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT1
Entrez Gene ID
23019
Gene Name
CCR4-NOT transcription complex subunit 1
Gene Aliases
AD-005, CDC39, NOT1, NOT1H
UniGene
Hs.460923
Gene Chromosome Location
Chr.16: 58519946-58629886 on Build GRCh38
Transcripts
NM_001265612.1 NM_016284.4 NR_049763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631377 c.6823C>T p.R2275W Chr.16: 58523464-58523464 on Build GRCh38
5628430 c.6817C>T p.Q2273* Chr.16: 58523470-58523470 on Build GRCh38
4505075 c.6838C>T p.H2280Y Chr.16: 58523449-58523449 on Build GRCh38
375630 c.6876T>C p.F2292F Chr.16: 58523411-58523411 on Build GRCh38
5754879 c.6888T>C p.N2296N Chr.16: 58523399-58523399 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771356837 Chr.16: 58523497-58523497 on Build GRCh38
rs941412039 Chr.16: 58523318-58523318 on Build GRCh38
rs76866497 Chr.16: 58523314-58523315 on Build GRCh38
rs11644694 Chr.16: 58523438-58523438 on Build GRCh38
rs888690435 Chr.16: 58523316-58523316 on Build GRCh38
rs758510366 Chr.16: 58523358-58523358 on Build GRCh38
rs199694259 Chr.16: 58523396-58523396 on Build GRCh38
rs760421587 Chr.16: 58523468-58523468 on Build GRCh38
rs768626343 Chr.16: 58523417-58523417 on Build GRCh38
rs934698042 Chr.16: 58523498-58523498 on Build GRCh38
rs772954031 Chr.16: 58523443-58523443 on Build GRCh38
rs968569086 Chr.16: 58523400-58523400 on Build GRCh38
rs180774956 Chr.16: 58523335-58523335 on Build GRCh38
rs755516483 Chr.16: 58523333-58523333 on Build GRCh38
rs576713015 Chr.16: 58523405-58523405 on Build GRCh38
rs398029533 Chr.16: 58523315-58523316 on Build GRCh38
rs752796604 Chr.16: 58523342-58523342 on Build GRCh38
rs150195298 Chr.16: 58523435-58523435 on Build GRCh38
rs1035641000 Chr.16: 58523315-58523316 on Build GRCh38
rs755845672 Chr.16: 58523372-58523372 on Build GRCh38
rs528353760 Chr.16: 58523364-58523364 on Build GRCh38
rs765740441 Chr.16: 58523528-58523528 on Build GRCh38
rs143651029 Chr.16: 58523360-58523360 on Build GRCh38
rs200707564 Chr.16: 58523363-58523363 on Build GRCh38
rs958176465 Chr.16: 58523496-58523496 on Build GRCh38
rs759954262 Chr.16: 58523380-58523380 on Build GRCh38
rs777053025 Chr.16: 58523519-58523519 on Build GRCh38
rs768250384 Chr.16: 58523423-58523423 on Build GRCh38
rs759864174 Chr.16: 58523526-58523526 on Build GRCh38
rs771740649 Chr.16: 58523441-58523441 on Build GRCh38

View Full Product Details