Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38782578-38782851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAL4
Entrez Gene ID
10126
Gene Name
dynein axonemal light chain 4
Gene Aliases
MRMV3, PIG27
UniGene
Hs.632766
Gene Chromosome Location
Chr.22: 38778508-38794156 on Build GRCh38
Transcripts
NM_005740.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6368139 c.29A>T p.E10V Chr.22: 38782703-38782703 on Build GRCh38
6855438 c.4G>T p.G2* Chr.22: 38782728-38782728 on Build GRCh38
4156270 c.36T>C p.D12D Chr.22: 38782696-38782696 on Build GRCh38
4156271 c.30G>T p.E10D Chr.22: 38782702-38782702 on Build GRCh38
3681435 c.36T>G p.D12E Chr.22: 38782696-38782696 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757824460 Chr.22: 38782763-38782763 on Build GRCh38
rs377441172 Chr.22: 38782618-38782618 on Build GRCh38
rs149945094 Chr.22: 38782812-38782812 on Build GRCh38
rs776374483 Chr.22: 38782700-38782700 on Build GRCh38
rs769563271 Chr.22: 38782712-38782712 on Build GRCh38
rs962259993 Chr.22: 38782697-38782697 on Build GRCh38
rs969392226 Chr.22: 38782687-38782687 on Build GRCh38
rs760271479 Chr.22: 38782766-38782766 on Build GRCh38
rs531184902 Chr.22: 38782657-38782657 on Build GRCh38
rs561379640 Chr.22: 38782757-38782757 on Build GRCh38
rs778365710 Chr.22: 38782682-38782682 on Build GRCh38
rs753061475 Chr.22: 38782751-38782751 on Build GRCh38
rs761779991 Chr.22: 38782624-38782624 on Build GRCh38
rs145196186 Chr.22: 38782688-38782688 on Build GRCh38
rs972326078 Chr.22: 38782773-38782773 on Build GRCh38
rs939816334 Chr.22: 38782752-38782752 on Build GRCh38
rs766536648 Chr.22: 38782720-38782720 on Build GRCh38
rs778851658 Chr.22: 38782640-38782641 on Build GRCh38
rs760482 Chr.22: 38782696-38782696 on Build GRCh38
rs755378290 Chr.22: 38782656-38782656 on Build GRCh38
rs768498769 Chr.22: 38782620-38782620 on Build GRCh38
rs759874004 Chr.22: 38782725-38782725 on Build GRCh38
rs188474370 Chr.22: 38782632-38782632 on Build GRCh38
rs547103096 Chr.22: 38782817-38782817 on Build GRCh38
rs764441010 Chr.22: 38782762-38782762 on Build GRCh38
rs373459643 Chr.22: 38782649-38782649 on Build GRCh38
rs150096420 Chr.22: 38782716-38782716 on Build GRCh38
rs773208580 Chr.22: 38782714-38782714 on Build GRCh38
rs560225069 Chr.22: 38782798-38782798 on Build GRCh38
rs750425135 Chr.22: 38782631-38782631 on Build GRCh38
rs765194725 Chr.22: 38782630-38782630 on Build GRCh38
rs776023216 Chr.22: 38782628-38782629 on Build GRCh38
rs775154267 Chr.22: 38782735-38782735 on Build GRCh38
rs376541254 Chr.22: 38782694-38782694 on Build GRCh38
rs776528001 Chr.22: 38782622-38782622 on Build GRCh38
rs147600184 Chr.22: 38782689-38782689 on Build GRCh38
rs761622559 Chr.22: 38782705-38782705 on Build GRCh38
rs141340064 Chr.22: 38782671-38782671 on Build GRCh38
rs563995135 Chr.22: 38782651-38782651 on Build GRCh38
rs762933968 Chr.22: 38782719-38782719 on Build GRCh38
rs138389147 Chr.22: 38782724-38782724 on Build GRCh38
rs767881922 Chr.22: 38782740-38782740 on Build GRCh38
rs779657592 Chr.22: 38782783-38782783 on Build GRCh38

View Full Product Details