Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 48544866-48545103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXB-AS1
Entrez Gene ID
100874362
Gene Name
HOXB cluster antisense RNA 1
Gene Aliases
-
UniGene
Hs.171806
Gene Chromosome Location
Chr.17: 48544351-48551241 on Build GRCh38
Transcripts
NR_102279.1 NM_002145.3

Gene Symbol
HOXB2
Entrez Gene ID
3212
Gene Name
homeobox B2
Gene Aliases
HOX2, HOX2H, Hox-2.8, K8
UniGene
Hs.514289
Gene Chromosome Location
Chr.17: 48542655-48545031 on Build GRCh38
Transcripts
NR_102279.1 NM_002145.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7115374 c.28G>A p.G10R Chr.17: 48544884-48544884 on Build GRCh38
6681672 c.25A>T p.I9F Chr.17: 48544887-48544887 on Build GRCh38
4844314 c.16G>T p.E6* Chr.17: 48544896-48544896 on Build GRCh38
5833324 c.27_28insT p.G10fs*156 Chr.17: 48544884-48544885 on Build GRCh38
5452850 c.38A>G p.N13S Chr.17: 48544874-48544874 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs199560772 Chr.17: 48544955-48544955 on Build GRCh38
rs370415244 Chr.17: 48544914-48544914 on Build GRCh38
rs960435015 Chr.17: 48544948-48544948 on Build GRCh38
rs879002160 Chr.17: 48544986-48544986 on Build GRCh38
rs946494054 Chr.17: 48544974-48544975 on Build GRCh38
rs138169880 Chr.17: 48544986-48544987 on Build GRCh38
rs544633760 Chr.17: 48544993-48544993 on Build GRCh38
rs779540329 Chr.17: 48544892-48544893 on Build GRCh38
rs933010621 Chr.17: 48545068-48545068 on Build GRCh38
rs988028925 Chr.17: 48544953-48544953 on Build GRCh38
rs146657238 Chr.17: 48544901-48544901 on Build GRCh38
rs756402530 Chr.17: 48544920-48544920 on Build GRCh38
rs886143588 Chr.17: 48544911-48544911 on Build GRCh38
rs979267693 Chr.17: 48544977-48544978 on Build GRCh38
rs779134754 Chr.17: 48544931-48544931 on Build GRCh38
rs746436357 Chr.17: 48544931-48544931 on Build GRCh38
rs754132459 Chr.17: 48544924-48544924 on Build GRCh38
rs780372151 Chr.17: 48544921-48544921 on Build GRCh38
rs9899351 Chr.17: 48544987-48544987 on Build GRCh38
rs368979011 Chr.17: 48545049-48545049 on Build GRCh38
rs747142697 Chr.17: 48544940-48544941 on Build GRCh38
rs901811939 Chr.17: 48545041-48545042 on Build GRCh38
rs529714338 Chr.17: 48544972-48544973 on Build GRCh38
rs748179497 Chr.17: 48544943-48544943 on Build GRCh38
rs111979968 Chr.17: 48544973-48544974 on Build GRCh38
rs922923334 Chr.17: 48545054-48545054 on Build GRCh38
rs780587153 Chr.17: 48544969-48544970 on Build GRCh38
rs913838894 Chr.17: 48544963-48544963 on Build GRCh38
rs772318292 Chr.17: 48544939-48544940 on Build GRCh38
rs760442320 Chr.17: 48545060-48545060 on Build GRCh38
rs866961469 Chr.17: 48544985-48544985 on Build GRCh38
rs533469950 Chr.17: 48545023-48545023 on Build GRCh38
rs111401880 Chr.17: 48545011-48545011 on Build GRCh38
rs555665958 Chr.17: 48544973-48544973 on Build GRCh38
rs775925985 Chr.17: 48544941-48544941 on Build GRCh38
rs925200314 Chr.17: 48545017-48545017 on Build GRCh38
rs1047925514 Chr.17: 48545059-48545059 on Build GRCh38
rs573064701 Chr.17: 48544925-48544925 on Build GRCh38
rs199712062 Chr.17: 48544983-48544984 on Build GRCh38

View Full Product Details