Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 49719711-49719952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM117A
Entrez Gene ID
81558
Gene Name
family with sequence similarity 117 member A
Gene Aliases
-
UniGene
Hs.514308
Gene Chromosome Location
Chr.17: 49710325-49764156 on Build GRCh38
Transcripts
NM_030802.3 XM_017025182.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4406470 c.671T>G p.V224G Chr.17: 49719797-49719797 on Build GRCh38
1740386 c.602C>T p.S201F Chr.17: 49719866-49719866 on Build GRCh38
1750124 c.705G>T p.L235L Chr.17: 49719763-49719763 on Build GRCh38
327592 c.575C>T p.A192V Chr.17: 49719893-49719893 on Build GRCh38
4770182 c.676delG p.V226fs*1 Chr.17: 49719792-49719792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757112053 Chr.17: 49719864-49719864 on Build GRCh38
rs773709536 Chr.17: 49719905-49719905 on Build GRCh38
rs967679314 Chr.17: 49719856-49719856 on Build GRCh38
rs749175258 Chr.17: 49719746-49719746 on Build GRCh38
rs747176001 Chr.17: 49719888-49719888 on Build GRCh38
rs199630851 Chr.17: 49719751-49719751 on Build GRCh38
rs763374167 Chr.17: 49719914-49719914 on Build GRCh38
rs765805148 Chr.17: 49719787-49719787 on Build GRCh38
rs374136431 Chr.17: 49719738-49719738 on Build GRCh38
rs964132705 Chr.17: 49719889-49719889 on Build GRCh38
rs368889183 Chr.17: 49719854-49719854 on Build GRCh38
rs767012921 Chr.17: 49719922-49719922 on Build GRCh38
rs576336246 Chr.17: 49719852-49719852 on Build GRCh38
rs772128488 Chr.17: 49719881-49719881 on Build GRCh38
rs780891419 Chr.17: 49719734-49719734 on Build GRCh38
rs199774395 Chr.17: 49719867-49719867 on Build GRCh38
rs777153106 Chr.17: 49719777-49719777 on Build GRCh38
rs745808831 Chr.17: 49719874-49719874 on Build GRCh38
rs770833755 Chr.17: 49719750-49719750 on Build GRCh38
rs536390470 Chr.17: 49719764-49719764 on Build GRCh38
rs762314978 Chr.17: 49719783-49719783 on Build GRCh38
rs201064183 Chr.17: 49719873-49719873 on Build GRCh38
rs768972931 Chr.17: 49719890-49719890 on Build GRCh38
rs780032318 Chr.17: 49719884-49719884 on Build GRCh38
rs750957462 Chr.17: 49719803-49719803 on Build GRCh38
rs752326587 Chr.17: 49719846-49719846 on Build GRCh38
rs777030465 Chr.17: 49719892-49719892 on Build GRCh38
rs566472842 Chr.17: 49719893-49719893 on Build GRCh38
rs116657589 Chr.17: 49719768-49719768 on Build GRCh38
rs759071955 Chr.17: 49719819-49719819 on Build GRCh38
rs983527112 Chr.17: 49719932-49719932 on Build GRCh38
rs772103873 Chr.17: 49719766-49719766 on Build GRCh38
rs368403545 Chr.17: 49719866-49719866 on Build GRCh38
rs755845613 Chr.17: 49719742-49719742 on Build GRCh38
rs769099323 Chr.17: 49719771-49719771 on Build GRCh38
rs762261590 Chr.17: 49719897-49719897 on Build GRCh38
rs766950126 Chr.17: 49719842-49719842 on Build GRCh38
rs753601420 Chr.17: 49719855-49719855 on Build GRCh38
rs748101996 Chr.17: 49719892-49719893 on Build GRCh38
rs760986356 Chr.17: 49719770-49719770 on Build GRCh38
rs770112797 Chr.17: 49719902-49719902 on Build GRCh38

View Full Product Details