Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 169380386-169380647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLZF1
Entrez Gene ID
8548
Gene Name
basic leucine zipper nuclear factor 1
Gene Aliases
GOLGIN-45, JEM-1, JEM-1s, JEM1
UniGene
Hs.130746
Gene Chromosome Location
Chr.1: 169367879-169396544 on Build GRCh38
Transcripts
NM_001320973.1 NM_003666.3 XM_005245561.4 XM_011510092.2 XM_017002645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7357574 c.778G>A p.E260K Chr.1: 169380590-169380590 on Build GRCh38
347937 c.729A>T p.A243A Chr.1: 169380541-169380541 on Build GRCh38
6476287 c.762G>T p.R254R Chr.1: 169380574-169380574 on Build GRCh38
6272791 c.676G>T p.A226S Chr.1: 169380488-169380488 on Build GRCh38
1728892 c.767A>T p.Q256L Chr.1: 169380579-169380579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780621408 Chr.1: 169380469-169380469 on Build GRCh38
rs768124390 Chr.1: 169380464-169380464 on Build GRCh38
rs148310730 Chr.1: 169380573-169380573 on Build GRCh38
rs900280942 Chr.1: 169380595-169380595 on Build GRCh38
rs759937276 Chr.1: 169380516-169380516 on Build GRCh38
rs544635266 Chr.1: 169380574-169380574 on Build GRCh38
rs760023199 Chr.1: 169380583-169380583 on Build GRCh38
rs778647086 Chr.1: 169380545-169380545 on Build GRCh38
rs774669127 Chr.1: 169380580-169380580 on Build GRCh38
rs771348947 Chr.1: 169380446-169380446 on Build GRCh38
rs750243416 Chr.1: 169380548-169380548 on Build GRCh38
rs756202911 Chr.1: 169380545-169380545 on Build GRCh38
rs746664112 Chr.1: 169380561-169380561 on Build GRCh38
rs138245508 Chr.1: 169380584-169380584 on Build GRCh38
rs527955070 Chr.1: 169380606-169380606 on Build GRCh38
rs144964235 Chr.1: 169380544-169380544 on Build GRCh38
rs368872376 Chr.1: 169380524-169380524 on Build GRCh38
rs769129174 Chr.1: 169380537-169380537 on Build GRCh38
rs113548186 Chr.1: 169380585-169380585 on Build GRCh38
rs779722099 Chr.1: 169380560-169380560 on Build GRCh38
rs779635588 Chr.1: 169380457-169380457 on Build GRCh38
rs765269804 Chr.1: 169380602-169380602 on Build GRCh38
rs766152591 Chr.1: 169380617-169380617 on Build GRCh38
rs1035484807 Chr.1: 169380541-169380541 on Build GRCh38
rs181614246 Chr.1: 169380462-169380462 on Build GRCh38
rs774785058 Chr.1: 169380505-169380505 on Build GRCh38
rs60461695 Chr.1: 169380549-169380549 on Build GRCh38
rs777784793 Chr.1: 169380560-169380562 on Build GRCh38
rs758168509 Chr.1: 169380554-169380554 on Build GRCh38
rs774681975 Chr.1: 169380572-169380572 on Build GRCh38
rs562436533 Chr.1: 169380525-169380525 on Build GRCh38
rs763506719 Chr.1: 169380472-169380472 on Build GRCh38
rs544422022 Chr.1: 169380496-169380496 on Build GRCh38
rs138335460 Chr.1: 169380521-169380521 on Build GRCh38
rs1012760625 Chr.1: 169380579-169380579 on Build GRCh38
rs758256718 Chr.1: 169380612-169380612 on Build GRCh38
rs761752160 Chr.1: 169380601-169380601 on Build GRCh38
rs139538765 Chr.1: 169380598-169380598 on Build GRCh38

View Full Product Details