Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 39988061-39988330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK13
Entrez Gene ID
8621
Gene Name
cyclin dependent kinase 13
Gene Aliases
CDC2L, CDC2L5, CHED, hCDK13
UniGene
Hs.233552
Gene Chromosome Location
Chr.7: 39949501-40097134 on Build GRCh38
Transcripts
NM_003718.4 NM_031267.3 XM_011515597.2 XM_017012750.1 XM_017012751.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1755272 c.1717A>T p.T573S Chr.7: 39988104-39988104 on Build GRCh38
3832760 c.1738C>G p.L580V Chr.7: 39988125-39988125 on Build GRCh38
6625375 c.1761T>C p.L587L Chr.7: 39988148-39988148 on Build GRCh38
4887229 c.1790A>G p.E597G Chr.7: 39988177-39988177 on Build GRCh38
5442203 c.1867G>C p.D623H Chr.7: 39988254-39988254 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773370535 Chr.7: 39988252-39988252 on Build GRCh38
rs914374352 Chr.7: 39988250-39988250 on Build GRCh38
rs55813129 Chr.7: 39988220-39988220 on Build GRCh38
rs1002791528 Chr.7: 39988125-39988125 on Build GRCh38
rs375662121 Chr.7: 39988264-39988264 on Build GRCh38
rs865901868 Chr.7: 39988108-39988108 on Build GRCh38
rs776004739 Chr.7: 39988089-39988089 on Build GRCh38
rs869175716 Chr.7: 39988289-39988289 on Build GRCh38
rs201843087 Chr.7: 39988302-39988302 on Build GRCh38
rs774684486 Chr.7: 39988105-39988105 on Build GRCh38
rs369518465 Chr.7: 39988106-39988106 on Build GRCh38
rs760696402 Chr.7: 39988255-39988255 on Build GRCh38
rs770369397 Chr.7: 39988087-39988087 on Build GRCh38
rs747897695 Chr.7: 39988168-39988168 on Build GRCh38
rs200341630 Chr.7: 39988213-39988213 on Build GRCh38
rs750412594 Chr.7: 39988112-39988112 on Build GRCh38
rs758076444 Chr.7: 39988283-39988283 on Build GRCh38
rs769104776 Chr.7: 39988205-39988205 on Build GRCh38
rs766263341 Chr.7: 39988123-39988123 on Build GRCh38
rs753653329 Chr.7: 39988145-39988145 on Build GRCh38
rs991811477 Chr.7: 39988263-39988263 on Build GRCh38
rs778536763 Chr.7: 39988158-39988158 on Build GRCh38
rs777254552 Chr.7: 39988287-39988287 on Build GRCh38
rs767581253 Chr.7: 39988096-39988098 on Build GRCh38
rs377402223 Chr.7: 39988184-39988184 on Build GRCh38
rs754814006 Chr.7: 39988150-39988150 on Build GRCh38
rs752381105 Chr.7: 39988282-39988282 on Build GRCh38
rs17496275 Chr.7: 39988257-39988257 on Build GRCh38
rs757977577 Chr.7: 39988173-39988173 on Build GRCh38
rs370048161 Chr.7: 39988266-39988266 on Build GRCh38
rs763370906 Chr.7: 39988096-39988096 on Build GRCh38
rs199654911 Chr.7: 39988183-39988183 on Build GRCh38
rs765140944 Chr.7: 39988279-39988279 on Build GRCh38
rs767693629 Chr.7: 39988107-39988107 on Build GRCh38
rs200811940 Chr.7: 39988098-39988098 on Build GRCh38
rs770457429 Chr.7: 39988197-39988197 on Build GRCh38
rs756047708 Chr.7: 39988113-39988113 on Build GRCh38
rs369158462 Chr.7: 39988214-39988214 on Build GRCh38
rs750898684 Chr.7: 39988121-39988121 on Build GRCh38
rs764436014 Chr.7: 39988103-39988103 on Build GRCh38
rs780659478 Chr.7: 39988199-39988199 on Build GRCh38
rs751137950 Chr.7: 39988295-39988295 on Build GRCh38

View Full Product Details