Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 102654260-102654503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARMCX5-GPRASP2
Entrez Gene ID
100528062
Gene Name
ARMCX5-GPRASP2 readthrough
Gene Aliases
-
UniGene
Hs.522729
Gene Chromosome Location
Chr.X: 102599348-102717733 on Build GRCh38
Transcripts
NM_001199818.1 NM_001099410.1 NM_001099411.1 NM_001184727.1 NM_014710.4 XM_017029981.1 XM_017029982.1

Gene Symbol
GPRASP1
Entrez Gene ID
9737
Gene Name
G protein-coupled receptor associated sorting protein 1
Gene Aliases
GASP, GASP-1, GASP1
UniGene
Hs.729265
Gene Chromosome Location
Chr.X: 102651366-102659083 on Build GRCh38
Transcripts
NM_001199818.1 NM_001099410.1 NM_001099411.1 NM_001184727.1 NM_014710.4 XM_017029981.1 XM_017029982.1

SNP Detail

SNP ID Chromosome Location
rs775068377 Chr.X: 102654431-102654431 on Build GRCh38
rs148368072 Chr.X: 102654292-102654292 on Build GRCh38
rs779157434 Chr.X: 102654385-102654385 on Build GRCh38
rs750370872 Chr.X: 102654320-102654320 on Build GRCh38
rs769320629 Chr.X: 102654302-102654302 on Build GRCh38
rs979793781 Chr.X: 102654364-102654364 on Build GRCh38
rs759901828 Chr.X: 102654454-102654454 on Build GRCh38
rs773994817 Chr.X: 102654467-102654467 on Build GRCh38
rs755177556 Chr.X: 102654375-102654375 on Build GRCh38
rs774833842 Chr.X: 102654464-102654466 on Build GRCh38
rs748183132 Chr.X: 102654388-102654388 on Build GRCh38
rs756717317 Chr.X: 102654315-102654315 on Build GRCh38
rs747514155 Chr.X: 102654413-102654413 on Build GRCh38
rs779963756 Chr.X: 102654337-102654337 on Build GRCh38
rs767572863 Chr.X: 102654316-102654316 on Build GRCh38
rs913569515 Chr.X: 102654442-102654442 on Build GRCh38
rs927034050 Chr.X: 102654451-102654451 on Build GRCh38
rs946338733 Chr.X: 102654448-102654448 on Build GRCh38
rs772134875 Chr.X: 102654392-102654392 on Build GRCh38
rs201355689 Chr.X: 102654473-102654473 on Build GRCh38
rs773769428 Chr.X: 102654399-102654399 on Build GRCh38
rs752970458 Chr.X: 102654334-102654334 on Build GRCh38
rs756235038 Chr.X: 102654423-102654423 on Build GRCh38
rs930351052 Chr.X: 102654471-102654471 on Build GRCh38
rs1023605044 Chr.X: 102654296-102654296 on Build GRCh38
rs188898027 Chr.X: 102654465-102654465 on Build GRCh38
rs749573637 Chr.X: 102654344-102654344 on Build GRCh38

View Full Product Details