Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 23523981-23524250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C18orf8
Entrez Gene ID
29919
Gene Name
chromosome 18 open reading frame 8
Gene Aliases
HsT2591, MIC1, Mic-1
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23503470-23531807 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_006722479.3 XM_011526015.1 XM_017025787.1

Gene Symbol
NPC1
Entrez Gene ID
4864
Gene Name
NPC intracellular cholesterol transporter 1
Gene Aliases
NPC
UniGene
Hs.464779
Gene Chromosome Location
Chr.18: 23506184-23586617 on Build GRCh38
Transcripts
NM_001276342.1 NM_001318707.1 NM_001318708.1 NM_001318709.1 NM_013326.4 XM_011525939.1 XM_017025717.1 XM_005258277.1 XM_005258279.2 XM_006722479.3 XM_011526015.1 XM_017025787.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5387884 c.986C>T p.S329F Chr.18: 23524154-23524154 on Build GRCh38
987077 c.991G>A p.V331I Chr.18: 23524159-23524159 on Build GRCh38
4444633 c.973G>A p.V325M Chr.18: 23524141-23524141 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs534203694 Chr.18: 23524035-23524035 on Build GRCh38
rs891385 Chr.18: 23524187-23524187 on Build GRCh38
rs766843454 Chr.18: 23524085-23524085 on Build GRCh38
rs979727154 Chr.18: 23524082-23524082 on Build GRCh38
rs752944530 Chr.18: 23524112-23524112 on Build GRCh38
rs891386 Chr.18: 23524007-23524007 on Build GRCh38
rs768120449 Chr.18: 23524178-23524178 on Build GRCh38
rs748824696 Chr.18: 23524177-23524177 on Build GRCh38
rs148467554 Chr.18: 23524186-23524186 on Build GRCh38
rs774042832 Chr.18: 23524179-23524179 on Build GRCh38
rs554160472 Chr.18: 23524121-23524121 on Build GRCh38
rs938103624 Chr.18: 23524202-23524202 on Build GRCh38
rs774004980 Chr.18: 23524227-23524228 on Build GRCh38
rs753032782 Chr.18: 23524215-23524215 on Build GRCh38
rs766817459 Chr.18: 23524197-23524197 on Build GRCh38
rs375981319 Chr.18: 23524099-23524099 on Build GRCh38
rs1024090418 Chr.18: 23524048-23524048 on Build GRCh38
rs763089805 Chr.18: 23524219-23524219 on Build GRCh38
rs969708209 Chr.18: 23524065-23524065 on Build GRCh38
rs773613606 Chr.18: 23524032-23524032 on Build GRCh38
rs772385916 Chr.18: 23524017-23524017 on Build GRCh38
rs761042685 Chr.18: 23524195-23524195 on Build GRCh38
rs765392851 Chr.18: 23524200-23524200 on Build GRCh38
rs531155366 Chr.18: 23524101-23524101 on Build GRCh38
rs755061519 Chr.18: 23524102-23524102 on Build GRCh38
rs780844044 Chr.18: 23524140-23524140 on Build GRCh38
rs370329557 Chr.18: 23524109-23524109 on Build GRCh38
rs143803533 Chr.18: 23524141-23524141 on Build GRCh38
rs747787855 Chr.18: 23524180-23524180 on Build GRCh38
rs572334098 Chr.18: 23524031-23524031 on Build GRCh38
rs201899092 Chr.18: 23524199-23524199 on Build GRCh38
rs371188093 Chr.18: 23524143-23524143 on Build GRCh38
rs777238935 Chr.18: 23524198-23524198 on Build GRCh38
rs746714905 Chr.18: 23524034-23524034 on Build GRCh38
rs779988837 Chr.18: 23524144-23524144 on Build GRCh38
rs765954710 Chr.18: 23524116-23524117 on Build GRCh38

View Full Product Details