Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 42552445-42552700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM19
Entrez Gene ID
114926
Gene Name
small integral membrane protein 19
Gene Aliases
C8orf40
UniGene
Hs.655320
Gene Chromosome Location
Chr.8: 42541155-42552997 on Build GRCh38
Transcripts
NM_001135674.1 NM_001135675.1 NM_001135676.1 NM_138436.3 XM_005273398.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
454592 c.277G>A p.D93N Chr.8: 42552582-42552582 on Build GRCh38
5718315 c.292C>T p.Q98* Chr.8: 42552576-42552576 on Build GRCh38
6180902 c.303A>G p.*101W Chr.8: 42552608-42552608 on Build GRCh38
3900303 c.271G>A p.D91N Chr.8: 42552555-42552555 on Build GRCh38
1489307 c.298G>A p.D100N Chr.8: 42552582-42552582 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138368316 Chr.8: 42552656-42552656 on Build GRCh38
rs143524150 Chr.8: 42552598-42552598 on Build GRCh38
rs368564087 Chr.8: 42552612-42552612 on Build GRCh38
rs751405372 Chr.8: 42552511-42552511 on Build GRCh38
rs866377307 Chr.8: 42552567-42552567 on Build GRCh38
rs746769055 Chr.8: 42552645-42552645 on Build GRCh38
rs746259533 Chr.8: 42552563-42552563 on Build GRCh38
rs759452198 Chr.8: 42552649-42552649 on Build GRCh38
rs370913111 Chr.8: 42552555-42552555 on Build GRCh38
rs975822970 Chr.8: 42552570-42552570 on Build GRCh38
rs754782206 Chr.8: 42552512-42552512 on Build GRCh38
rs751495430 Chr.8: 42552644-42552644 on Build GRCh38
rs766452576 Chr.8: 42552637-42552637 on Build GRCh38
rs747811914 Chr.8: 42552520-42552520 on Build GRCh38
rs766362399 Chr.8: 42552506-42552506 on Build GRCh38
rs775617744 Chr.8: 42552589-42552589 on Build GRCh38
rs532252880 Chr.8: 42552559-42552559 on Build GRCh38
rs906033745 Chr.8: 42552603-42552603 on Build GRCh38
rs375904934 Chr.8: 42552539-42552539 on Build GRCh38
rs1057364933 Chr.8: 42552653-42552653 on Build GRCh38
rs755619011 Chr.8: 42552526-42552526 on Build GRCh38
rs771436968 Chr.8: 42552554-42552554 on Build GRCh38
rs781669909 Chr.8: 42552519-42552519 on Build GRCh38
rs761918301 Chr.8: 42552632-42552632 on Build GRCh38
rs986239444 Chr.8: 42552517-42552517 on Build GRCh38
rs772407105 Chr.8: 42552568-42552568 on Build GRCh38
rs930175126 Chr.8: 42552623-42552623 on Build GRCh38
rs776744373 Chr.8: 42552508-42552511 on Build GRCh38
rs1032956162 Chr.8: 42552503-42552503 on Build GRCh38
rs749848286 Chr.8: 42552540-42552540 on Build GRCh38
rs760951604 Chr.8: 42552597-42552597 on Build GRCh38
rs765406132 Chr.8: 42552594-42552595 on Build GRCh38
rs941788880 Chr.8: 42552536-42552536 on Build GRCh38
rs1017263520 Chr.8: 42552521-42552521 on Build GRCh38
rs780729392 Chr.8: 42552513-42552513 on Build GRCh38
rs759771092 Chr.8: 42552531-42552533 on Build GRCh38

View Full Product Details