Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 91756252-91756515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPC5
Entrez Gene ID
2262
Gene Name
glypican 5
Gene Aliases
-
UniGene
Hs.655675
Gene Chromosome Location
Chr.13: 91398619-92867237 on Build GRCh38
Transcripts
NM_004466.5 XM_011521054.2 XM_011521055.2 XM_011521056.2 XM_011521057.2 XM_011521058.2 XM_011521059.2 XM_011521060.2 XM_017020435.1 XM_017020436.1 XM_017020437.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6842957 c.1155-2A>G p.? Chr.13: 91756293-91756293 on Build GRCh38
4395955 c.1225G>A p.E409K Chr.13: 91756365-91756365 on Build GRCh38
6673186 c.1198G>A p.G400S Chr.13: 91756338-91756338 on Build GRCh38
3469944 c.1250C>T p.P417L Chr.13: 91756390-91756390 on Build GRCh38
432610 c.1197A>C p.G399G Chr.13: 91756337-91756337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750057478 Chr.13: 91756450-91756450 on Build GRCh38
rs769996318 Chr.13: 91756397-91756397 on Build GRCh38
rs748659135 Chr.13: 91756364-91756364 on Build GRCh38
rs138163539 Chr.13: 91756381-91756381 on Build GRCh38
rs778685770 Chr.13: 91756285-91756285 on Build GRCh38
rs779335662 Chr.13: 91756363-91756363 on Build GRCh38
rs368890147 Chr.13: 91756409-91756409 on Build GRCh38
rs762969147 Chr.13: 91756408-91756408 on Build GRCh38
rs1000475109 Chr.13: 91756321-91756321 on Build GRCh38
rs375902771 Chr.13: 91756291-91756291 on Build GRCh38
rs970853421 Chr.13: 91756425-91756425 on Build GRCh38
rs781056098 Chr.13: 91756468-91756468 on Build GRCh38
rs772637603 Chr.13: 91756369-91756369 on Build GRCh38
rs529941640 Chr.13: 91756333-91756333 on Build GRCh38
rs755603177 Chr.13: 91756350-91756350 on Build GRCh38
rs775775860 Chr.13: 91756400-91756400 on Build GRCh38
rs767150002 Chr.13: 91756447-91756447 on Build GRCh38
rs904679286 Chr.13: 91756307-91756307 on Build GRCh38
rs144633839 Chr.13: 91756342-91756342 on Build GRCh38
rs750058767 Chr.13: 91756474-91756474 on Build GRCh38
rs776737101 Chr.13: 91756390-91756390 on Build GRCh38
rs771156014 Chr.13: 91756383-91756383 on Build GRCh38
rs757639727 Chr.13: 91756384-91756384 on Build GRCh38
rs756910046 Chr.13: 91756337-91756337 on Build GRCh38
rs760951494 Chr.13: 91756289-91756289 on Build GRCh38
rs915980144 Chr.13: 91756314-91756314 on Build GRCh38
rs201682963 Chr.13: 91756372-91756372 on Build GRCh38
rs752691145 Chr.13: 91756322-91756322 on Build GRCh38
rs761543755 Chr.13: 91756430-91756430 on Build GRCh38
rs759578084 Chr.13: 91756309-91756309 on Build GRCh38
rs776752383 Chr.13: 91756308-91756308 on Build GRCh38
rs1040910171 Chr.13: 91756482-91756482 on Build GRCh38
rs765214865 Chr.13: 91756315-91756315 on Build GRCh38
rs778271177 Chr.13: 91756310-91756310 on Build GRCh38
rs575206298 Chr.13: 91756348-91756348 on Build GRCh38
rs1009196812 Chr.13: 91756346-91756346 on Build GRCh38
rs759598195 Chr.13: 91756391-91756391 on Build GRCh38
rs762766112 Chr.13: 91756330-91756330 on Build GRCh38
rs751337297 Chr.13: 91756420-91756420 on Build GRCh38
rs755556515 Chr.13: 91756470-91756470 on Build GRCh38

View Full Product Details