Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 10703972-10704199 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YBX3
Entrez Gene ID
8531
Gene Name
Y-box binding protein 3
Gene Aliases
CSDA, CSDA1, DBPA, ZONAB
UniGene
Hs.221889
Gene Chromosome Location
Chr.12: 10699077-10723448 on Build GRCh38
Transcripts
NM_001145426.1 NM_003651.4 XM_017020122.1 XM_017020123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3870654 c.873C>T p.Y291Y Chr.12: 10704056-10704056 on Build GRCh38
5438603 c.842T>G p.V281G Chr.12: 10704087-10704087 on Build GRCh38
5970485 c.794G>A p.G265E Chr.12: 10704135-10704135 on Build GRCh38
319687 c.824C>T p.A275V Chr.12: 10704105-10704105 on Build GRCh38
5940248 c.781-4C>T p.? Chr.12: 10704152-10704152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148757533 Chr.12: 10704081-10704081 on Build GRCh38
rs372644769 Chr.12: 10704157-10704157 on Build GRCh38
rs956389688 Chr.12: 10704137-10704137 on Build GRCh38
rs746683020 Chr.12: 10704061-10704061 on Build GRCh38
rs756480754 Chr.12: 10704093-10704093 on Build GRCh38
rs181901839 Chr.12: 10704046-10704046 on Build GRCh38
rs757865246 Chr.12: 10704055-10704055 on Build GRCh38
rs777444749 Chr.12: 10704059-10704059 on Build GRCh38
rs777199498 Chr.12: 10704144-10704144 on Build GRCh38
rs775779889 Chr.12: 10704013-10704013 on Build GRCh38
rs773232649 Chr.12: 10704110-10704110 on Build GRCh38
rs776681989 Chr.12: 10704002-10704002 on Build GRCh38
rs114697298 Chr.12: 10704054-10704054 on Build GRCh38
rs111997295 Chr.12: 10704151-10704151 on Build GRCh38
rs371991259 Chr.12: 10704089-10704089 on Build GRCh38
rs776629580 Chr.12: 10704065-10704065 on Build GRCh38
rs113414468 Chr.12: 10704049-10704049 on Build GRCh38
rs528288967 Chr.12: 10704171-10704171 on Build GRCh38
rs371675251 Chr.12: 10704178-10704178 on Build GRCh38
rs759580471 Chr.12: 10704043-10704043 on Build GRCh38
rs754162494 Chr.12: 10704037-10704037 on Build GRCh38
rs141479423 Chr.12: 10704090-10704090 on Build GRCh38
rs769787844 Chr.12: 10704067-10704067 on Build GRCh38
rs898264692 Chr.12: 10704034-10704035 on Build GRCh38
rs111494814 Chr.12: 10704149-10704149 on Build GRCh38
rs754330079 Chr.12: 10704111-10704111 on Build GRCh38
rs780996817 Chr.12: 10704173-10704173 on Build GRCh38
rs777107841 Chr.12: 10704048-10704048 on Build GRCh38
rs756931343 Chr.12: 10704162-10704162 on Build GRCh38
rs375433594 Chr.12: 10704135-10704135 on Build GRCh38
rs764322042 Chr.12: 10704031-10704031 on Build GRCh38
rs762946283 Chr.12: 10704028-10704028 on Build GRCh38
rs770666557 Chr.12: 10704000-10704000 on Build GRCh38
rs745797213 Chr.12: 10704066-10704066 on Build GRCh38
rs114117963 Chr.12: 10704073-10704073 on Build GRCh38
rs764791914 Chr.12: 10704110-10704111 on Build GRCh38
rs769736570 Chr.12: 10704177-10704177 on Build GRCh38
rs976660132 Chr.12: 10704133-10704133 on Build GRCh38
rs757609940 Chr.12: 10704034-10704034 on Build GRCh38
rs993631790 Chr.12: 10704060-10704060 on Build GRCh38
rs759599637 Chr.12: 10704004-10704004 on Build GRCh38
rs745601237 Chr.12: 10704176-10704176 on Build GRCh38
rs770741003 Chr.12: 10704063-10704063 on Build GRCh38
rs765398668 Chr.12: 10704010-10704010 on Build GRCh38
rs766820092 Chr.12: 10704101-10704101 on Build GRCh38
rs376972136 Chr.12: 10704033-10704033 on Build GRCh38
rs764118341 Chr.12: 10704076-10704076 on Build GRCh38
rs755332920 Chr.12: 10704054-10704055 on Build GRCh38
rs368389577 Chr.12: 10704044-10704044 on Build GRCh38

View Full Product Details