Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32867688-32867923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNDC5
Entrez Gene ID
252995
Gene Name
fibronectin type III domain containing 5
Gene Aliases
FRCP2, irisin
UniGene
Hs.524234
Gene Chromosome Location
Chr.1: 32862268-32872492 on Build GRCh38
Transcripts
NM_001171940.1 NM_001171941.2 NM_153756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3865338 c.198G>C p.M66I Chr.1: 32867829-32867829 on Build GRCh38
6587458 c.235G>A p.E79K Chr.1: 32867792-32867792 on Build GRCh38
4539624 c.270G>A p.W90* Chr.1: 32867757-32867757 on Build GRCh38
6665916 c.242T>C p.L81P Chr.1: 32867785-32867785 on Build GRCh38
908364 c.202G>T p.E68* Chr.1: 32867825-32867825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757750681 Chr.1: 32867829-32867829 on Build GRCh38
rs539108566 Chr.1: 32867894-32867894 on Build GRCh38
rs985805949 Chr.1: 32867846-32867846 on Build GRCh38
rs771678427 Chr.1: 32867762-32867762 on Build GRCh38
rs369352278 Chr.1: 32867733-32867733 on Build GRCh38
rs777889321 Chr.1: 32867778-32867780 on Build GRCh38
rs778363778 Chr.1: 32867737-32867737 on Build GRCh38
rs189231805 Chr.1: 32867777-32867777 on Build GRCh38
rs367966100 Chr.1: 32867738-32867738 on Build GRCh38
rs137888693 Chr.1: 32867800-32867800 on Build GRCh38
rs773770710 Chr.1: 32867755-32867755 on Build GRCh38
rs138727728 Chr.1: 32867786-32867786 on Build GRCh38
rs373703377 Chr.1: 32867852-32867852 on Build GRCh38
rs770424395 Chr.1: 32867735-32867735 on Build GRCh38
rs370205345 Chr.1: 32867809-32867809 on Build GRCh38
rs768402369 Chr.1: 32867889-32867889 on Build GRCh38
rs755217107 Chr.1: 32867793-32867793 on Build GRCh38
rs555122724 Chr.1: 32867903-32867903 on Build GRCh38
rs756182862 Chr.1: 32867723-32867725 on Build GRCh38
rs776502849 Chr.1: 32867778-32867778 on Build GRCh38
rs745434931 Chr.1: 32867840-32867840 on Build GRCh38
rs774389746 Chr.1: 32867866-32867867 on Build GRCh38
rs751766890 Chr.1: 32867712-32867712 on Build GRCh38
rs781583154 Chr.1: 32867872-32867872 on Build GRCh38
rs755197969 Chr.1: 32867716-32867716 on Build GRCh38
rs775260802 Chr.1: 32867772-32867772 on Build GRCh38
rs746686746 Chr.1: 32867885-32867885 on Build GRCh38
rs568822407 Chr.1: 32867849-32867849 on Build GRCh38
rs145120852 Chr.1: 32867719-32867719 on Build GRCh38
rs749367886 Chr.1: 32867835-32867836 on Build GRCh38
rs545975920 Chr.1: 32867849-32867850 on Build GRCh38
rs778471724 Chr.1: 32867838-32867838 on Build GRCh38
rs749779922 Chr.1: 32867828-32867828 on Build GRCh38
rs113173936 Chr.1: 32867896-32867896 on Build GRCh38
rs371687782 Chr.1: 32867865-32867865 on Build GRCh38
rs770432237 Chr.1: 32867744-32867744 on Build GRCh38
rs370217730 Chr.1: 32867857-32867858 on Build GRCh38
rs1029013109 Chr.1: 32867818-32867818 on Build GRCh38
rs377741902 Chr.1: 32867803-32867803 on Build GRCh38
rs775064555 Chr.1: 32867881-32867881 on Build GRCh38
rs981566156 Chr.1: 32867726-32867726 on Build GRCh38
rs371838288 Chr.1: 32867877-32867877 on Build GRCh38
rs745510759 Chr.1: 32867760-32867760 on Build GRCh38
rs763891316 Chr.1: 32867775-32867775 on Build GRCh38
rs557583773 Chr.1: 32867895-32867895 on Build GRCh38
rs748586202 Chr.1: 32867734-32867734 on Build GRCh38
rs893902374 Chr.1: 32867780-32867780 on Build GRCh38
rs963125076 Chr.1: 32867892-32867892 on Build GRCh38
rs761689081 Chr.1: 32867781-32867781 on Build GRCh38
rs766452920 Chr.1: 32867784-32867784 on Build GRCh38
rs533703704 Chr.1: 32867901-32867901 on Build GRCh38
rs200824057 Chr.1: 32867774-32867774 on Build GRCh38
rs771615717 Chr.1: 32867864-32867864 on Build GRCh38

View Full Product Details