Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 56315322-56315547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211
Entrez Gene ID
57482
Gene Name
KIAA1211
Gene Aliases
-
UniGene
Hs.596667
Gene Chromosome Location
Chr.4: 56170195-56330724 on Build GRCh38
Transcripts
NM_020722.1 XM_005265752.2 XM_005265755.2 XM_005265756.2 XM_006714045.2 XM_011534397.2 XM_017008473.1 XM_017008474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4773402 c.1984G>A p.A662T Chr.4: 56315486-56315486 on Build GRCh38
5655987 c.2014C>T p.R672W Chr.4: 56315516-56315516 on Build GRCh38
7091677 c.2038G>A p.E680K Chr.4: 56315540-56315540 on Build GRCh38
4516425 c.1912_1913CC>TT p.P638F Chr.4: 56315414-56315415 on Build GRCh38
4125261 c.1919G>A p.R640Q Chr.4: 56315421-56315421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777771925 Chr.4: 56315488-56315488 on Build GRCh38
rs533055150 Chr.4: 56315468-56315468 on Build GRCh38
rs752102687 Chr.4: 56315431-56315431 on Build GRCh38
rs202151867 Chr.4: 56315384-56315384 on Build GRCh38
rs764620167 Chr.4: 56315429-56315429 on Build GRCh38
rs755661717 Chr.4: 56315442-56315442 on Build GRCh38
rs549120782 Chr.4: 56315467-56315467 on Build GRCh38
rs778638578 Chr.4: 56315484-56315484 on Build GRCh38
rs201547934 Chr.4: 56315392-56315392 on Build GRCh38
rs190006805 Chr.4: 56315491-56315491 on Build GRCh38
rs748045935 Chr.4: 56315450-56315450 on Build GRCh38
rs527908970 Chr.4: 56315437-56315437 on Build GRCh38
rs765860702 Chr.4: 56315471-56315471 on Build GRCh38
rs774263670 Chr.4: 56315373-56315373 on Build GRCh38
rs753503661 Chr.4: 56315479-56315479 on Build GRCh38
rs965392934 Chr.4: 56315355-56315355 on Build GRCh38
rs775482840 Chr.4: 56315507-56315507 on Build GRCh38
rs769529519 Chr.4: 56315363-56315363 on Build GRCh38
rs749119312 Chr.4: 56315367-56315367 on Build GRCh38
rs934818191 Chr.4: 56315410-56315410 on Build GRCh38
rs779759747 Chr.4: 56315443-56315443 on Build GRCh38
rs1044854436 Chr.4: 56315521-56315521 on Build GRCh38
rs958009471 Chr.4: 56315356-56315356 on Build GRCh38
rs748955966 Chr.4: 56315444-56315444 on Build GRCh38
rs765516988 Chr.4: 56315399-56315399 on Build GRCh38
rs758272933 Chr.4: 56315486-56315486 on Build GRCh38
rs749983086 Chr.4: 56315439-56315439 on Build GRCh38
rs368631356 Chr.4: 56315366-56315366 on Build GRCh38
rs769843830 Chr.4: 56315461-56315461 on Build GRCh38
rs752947513 Chr.4: 56315405-56315405 on Build GRCh38
rs372605669 Chr.4: 56315485-56315485 on Build GRCh38
rs749494821 Chr.4: 56315527-56315527 on Build GRCh38
rs7672073 Chr.4: 56315466-56315466 on Build GRCh38
rs776522422 Chr.4: 56315457-56315457 on Build GRCh38
rs754664789 Chr.4: 56315483-56315483 on Build GRCh38
rs768422976 Chr.4: 56315446-56315446 on Build GRCh38
rs778728205 Chr.4: 56315448-56315448 on Build GRCh38
rs61750793 Chr.4: 56315463-56315463 on Build GRCh38
rs886146243 Chr.4: 56315475-56315475 on Build GRCh38
rs768408331 Chr.4: 56315370-56315370 on Build GRCh38
rs763382073 Chr.4: 56315412-56315412 on Build GRCh38
rs759635335 Chr.4: 56315398-56315398 on Build GRCh38
rs770975009 Chr.4: 56315391-56315391 on Build GRCh38
rs769175297 Chr.4: 56315461-56315462 on Build GRCh38
rs1054857144 Chr.4: 56315419-56315419 on Build GRCh38
rs759675795 Chr.4: 56315459-56315459 on Build GRCh38
rs769904803 Chr.4: 56315494-56315494 on Build GRCh38
rs989454160 Chr.4: 56315362-56315362 on Build GRCh38

View Full Product Details