Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77622425-77622660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLNS1A
Entrez Gene ID
1207
Gene Name
chloride nucleotide-sensitive channel 1A
Gene Aliases
CLCI, CLNS1B, ICln
UniGene
Hs.430733
Gene Chromosome Location
Chr.11: 77616151-77637806 on Build GRCh38
Transcripts
NM_001293.2 NM_001311199.1 NM_001311200.1 NM_001311201.1 NM_001311202.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1628233 c.631A>G p.I211V Chr.11: 77622515-77622515 on Build GRCh38
4898119 c.518G>A p.G173E Chr.11: 77622628-77622628 on Build GRCh38
1561968 c.558G>T p.E186D Chr.11: 77622588-77622588 on Build GRCh38
7183151 c.524C>T p.S175F Chr.11: 77622622-77622622 on Build GRCh38
4757782 c.506C>T p.T169I Chr.11: 77622640-77622640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752972357 Chr.11: 77622541-77622542 on Build GRCh38
rs546073378 Chr.11: 77622510-77622510 on Build GRCh38
rs942524212 Chr.11: 77622575-77622575 on Build GRCh38
rs761676085 Chr.11: 77622619-77622619 on Build GRCh38
rs200217752 Chr.11: 77622458-77622458 on Build GRCh38
rs965787768 Chr.11: 77622603-77622603 on Build GRCh38
rs375032831 Chr.11: 77622469-77622469 on Build GRCh38
rs767917208 Chr.11: 77622480-77622480 on Build GRCh38
rs750932416 Chr.11: 77622487-77622487 on Build GRCh38
rs144656096 Chr.11: 77622605-77622605 on Build GRCh38
rs1038623370 Chr.11: 77622583-77622583 on Build GRCh38
rs777595695 Chr.11: 77622556-77622556 on Build GRCh38
rs758198126 Chr.11: 77622550-77622550 on Build GRCh38
rs756618849 Chr.11: 77622490-77622490 on Build GRCh38
rs762142514 Chr.11: 77622474-77622474 on Build GRCh38
rs770943692 Chr.11: 77622577-77622577 on Build GRCh38
rs779053169 Chr.11: 77622519-77622519 on Build GRCh38
rs746971008 Chr.11: 77622571-77622571 on Build GRCh38
rs769414519 Chr.11: 77622607-77622607 on Build GRCh38
rs753870233 Chr.11: 77622499-77622499 on Build GRCh38
rs780128165 Chr.11: 77622494-77622494 on Build GRCh38
rs756396777 Chr.11: 77622623-77622625 on Build GRCh38
rs748390965 Chr.11: 77622532-77622532 on Build GRCh38
rs889605965 Chr.11: 77622537-77622537 on Build GRCh38
rs775069705 Chr.11: 77622613-77622613 on Build GRCh38

View Full Product Details