Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136412755-136413004 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PMPCA
Entrez Gene ID
23203
Gene Name
peptidase, mitochondrial processing alpha subunit
Gene Aliases
Alpha-MPP, INPP5E, P-55, SCAR2
UniGene
Hs.495471
Gene Chromosome Location
Chr.9: 136410573-136423761 on Build GRCh38
Transcripts
NM_001282944.1 NM_001282946.1 NM_015160.2 XM_005266059.4 XM_017014543.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4801423 c.407A>G p.H136R Chr.9: 136412862-136412862 on Build GRCh38
1106782 c.362C>T p.A121V Chr.9: 136412817-136412817 on Build GRCh38
4747927 c.418_419delTG p.C140fs*1 Chr.9: 136412873-136412874 on Build GRCh38
1701961 c.383A>T p.E128V Chr.9: 136412838-136412838 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs935843834 Chr.9: 136412945-136412945 on Build GRCh38
rs776763580 Chr.9: 136412917-136412917 on Build GRCh38
rs371727062 Chr.9: 136412929-136412929 on Build GRCh38
rs767701205 Chr.9: 136412778-136412778 on Build GRCh38
rs370502909 Chr.9: 136412852-136412852 on Build GRCh38
rs187951510 Chr.9: 136412819-136412819 on Build GRCh38
rs763420915 Chr.9: 136412861-136412861 on Build GRCh38
rs753883958 Chr.9: 136412788-136412788 on Build GRCh38
rs200019750 Chr.9: 136412863-136412863 on Build GRCh38
rs760797809 Chr.9: 136412905-136412905 on Build GRCh38
rs149377026 Chr.9: 136412927-136412927 on Build GRCh38
rs752569851 Chr.9: 136412816-136412816 on Build GRCh38
rs112738288 Chr.9: 136412982-136412982 on Build GRCh38
rs770498809 Chr.9: 136412829-136412829 on Build GRCh38
rs1054286787 Chr.9: 136412951-136412951 on Build GRCh38
rs922553423 Chr.9: 136412893-136412893 on Build GRCh38
rs558316725 Chr.9: 136412942-136412942 on Build GRCh38
rs761936198 Chr.9: 136412795-136412795 on Build GRCh38
rs777428813 Chr.9: 136412932-136412932 on Build GRCh38
rs767789432 Chr.9: 136412887-136412887 on Build GRCh38
rs780779529 Chr.9: 136412941-136412941 on Build GRCh38
rs777534357 Chr.9: 136412820-136412820 on Build GRCh38
rs778649107 Chr.9: 136412811-136412811 on Build GRCh38
rs751273505 Chr.9: 136412934-136412934 on Build GRCh38
rs961600848 Chr.9: 136412825-136412825 on Build GRCh38
rs750536367 Chr.9: 136412901-136412901 on Build GRCh38
rs780671064 Chr.9: 136412843-136412843 on Build GRCh38
rs770360676 Chr.9: 136412851-136412851 on Build GRCh38
rs898032054 Chr.9: 136412792-136412792 on Build GRCh38
rs774779672 Chr.9: 136412866-136412866 on Build GRCh38
rs754979744 Chr.9: 136412793-136412793 on Build GRCh38
rs377406908 Chr.9: 136412850-136412850 on Build GRCh38
rs891725940 Chr.9: 136412973-136412973 on Build GRCh38
rs762112786 Chr.9: 136412868-136412868 on Build GRCh38
rs374699145 Chr.9: 136412936-136412936 on Build GRCh38
rs375358633 Chr.9: 136412898-136412898 on Build GRCh38
rs910175313 Chr.9: 136412795-136412795 on Build GRCh38

View Full Product Details