Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11397437-11397664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGL3
Entrez Gene ID
57139
Gene Name
ral guanine nucleotide dissociation stimulator like 3
Gene Aliases
-
UniGene
Hs.375142
Gene Chromosome Location
Chr.19: 11394056-11419342 on Build GRCh38
Transcripts
NM_001035223.3 NM_001161616.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5725154 c.1852C>T p.R618C Chr.19: 11397492-11397492 on Build GRCh38
7256487 c.1848C>T p.V616V Chr.19: 11397496-11397496 on Build GRCh38
3718065 c.1849A>T p.S617C Chr.19: 11397513-11397513 on Build GRCh38
3718066 c.1831A>T p.S611C Chr.19: 11397513-11397513 on Build GRCh38
6381176 c.1790G>T p.S597I Chr.19: 11397572-11397572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896975362 Chr.19: 11397461-11397461 on Build GRCh38
rs759428729 Chr.19: 11397564-11397564 on Build GRCh38
rs759030377 Chr.19: 11397486-11397486 on Build GRCh38
rs750186888 Chr.19: 11397619-11397629 on Build GRCh38
rs781284504 Chr.19: 11397495-11397495 on Build GRCh38
rs139553006 Chr.19: 11397468-11397468 on Build GRCh38
rs575778869 Chr.19: 11397623-11397623 on Build GRCh38
rs978938242 Chr.19: 11397525-11397525 on Build GRCh38
rs201088422 Chr.19: 11397592-11397592 on Build GRCh38
rs201757947 Chr.19: 11397490-11397490 on Build GRCh38
rs536764443 Chr.19: 11397509-11397509 on Build GRCh38
rs770782869 Chr.19: 11397641-11397641 on Build GRCh38
rs779182992 Chr.19: 11397475-11397475 on Build GRCh38
rs777971255 Chr.19: 11397542-11397542 on Build GRCh38
rs139444484 Chr.19: 11397520-11397520 on Build GRCh38
rs774088081 Chr.19: 11397558-11397558 on Build GRCh38
rs770731388 Chr.19: 11397557-11397557 on Build GRCh38
rs201609417 Chr.19: 11397523-11397523 on Build GRCh38
rs753557516 Chr.19: 11397585-11397585 on Build GRCh38
rs769971185 Chr.19: 11397541-11397541 on Build GRCh38
rs752133506 Chr.19: 11397527-11397527 on Build GRCh38
rs2291516 Chr.19: 11397501-11397501 on Build GRCh38
rs376798441 Chr.19: 11397508-11397508 on Build GRCh38
rs755747168 Chr.19: 11397469-11397469 on Build GRCh38
rs557601517 Chr.19: 11397593-11397593 on Build GRCh38
rs770029288 Chr.19: 11397500-11397500 on Build GRCh38
rs946032851 Chr.19: 11397637-11397637 on Build GRCh38
rs763630083 Chr.19: 11397573-11397573 on Build GRCh38
rs879183341 Chr.19: 11397494-11397494 on Build GRCh38
rs755488135 Chr.19: 11397602-11397602 on Build GRCh38
rs371910024 Chr.19: 11397524-11397524 on Build GRCh38
rs762725979 Chr.19: 11397505-11397505 on Build GRCh38
rs1001169516 Chr.19: 11397492-11397492 on Build GRCh38
rs1024065504 Chr.19: 11397544-11397544 on Build GRCh38
rs772020574 Chr.19: 11397566-11397566 on Build GRCh38
rs568216829 Chr.19: 11397571-11397571 on Build GRCh38
rs111956126 Chr.19: 11397516-11397516 on Build GRCh38
rs751922294 Chr.19: 11397594-11397594 on Build GRCh38
rs775380798 Chr.19: 11397617-11397617 on Build GRCh38
rs749448742 Chr.19: 11397545-11397545 on Build GRCh38
rs1036431465 Chr.19: 11397603-11397603 on Build GRCh38
rs552123333 Chr.19: 11397512-11397512 on Build GRCh38
rs766988770 Chr.19: 11397464-11397464 on Build GRCh38
rs780700264 Chr.19: 11397489-11397489 on Build GRCh38
rs267605283 Chr.19: 11397536-11397536 on Build GRCh38
rs750883806 Chr.19: 11397521-11397521 on Build GRCh38
rs747582026 Chr.19: 11397480-11397480 on Build GRCh38
rs971587170 Chr.19: 11397553-11397553 on Build GRCh38
rs746209386 Chr.19: 11397476-11397476 on Build GRCh38
rs374802887 Chr.19: 11397531-11397531 on Build GRCh38
rs201152952 Chr.19: 11397456-11397456 on Build GRCh38
rs756194461 Chr.19: 11397621-11397621 on Build GRCh38
rs777776627 Chr.19: 11397634-11397634 on Build GRCh38
rs147520409 Chr.19: 11397458-11397458 on Build GRCh38
rs748237597 Chr.19: 11397537-11397537 on Build GRCh38
rs761507731 Chr.19: 11397587-11397587 on Build GRCh38
rs774918397 Chr.19: 11397567-11397567 on Build GRCh38
rs367697602 Chr.19: 11397580-11397580 on Build GRCh38
rs201346905 Chr.19: 11397491-11397491 on Build GRCh38
rs575260383 Chr.19: 11397589-11397589 on Build GRCh38
rs926199415 Chr.19: 11397530-11397530 on Build GRCh38
rs781716689 Chr.19: 11397607-11397607 on Build GRCh38

View Full Product Details