Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6977554-6977793 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
UniGene
Hs.558447
Gene Chromosome Location
Chr.12: 6970781-6997428 on Build GRCh38
Transcripts
NR_135131.1 NM_005768.5

Gene Symbol
LPCAT3
Entrez Gene ID
10162
Gene Name
lysophosphatidylcholine acyltransferase 3
Gene Aliases
C3F, LPCAT, LPLAT 5, LPSAT, MBOAT5, OACT5, nessy
UniGene
Hs.655248
Gene Chromosome Location
Chr.12: 6976185-7018538 on Build GRCh38
Transcripts
NR_135131.1 NM_005768.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
234923 c.1179G>A p.V393V Chr.12: 6977607-6977607 on Build GRCh38
7142888 c.1055G>T p.R352L Chr.12: 6977731-6977731 on Build GRCh38
6702328 c.1042T>C p.Y348H Chr.12: 6977744-6977744 on Build GRCh38
2100502 c.1122C>T p.H374H Chr.12: 6977664-6977664 on Build GRCh38
4698445 c.1102C>T p.L368L Chr.12: 6977684-6977684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782278519 Chr.12: 6977725-6977725 on Build GRCh38
rs782677899 Chr.12: 6977756-6977756 on Build GRCh38
rs376008049 Chr.12: 6977731-6977731 on Build GRCh38
rs782736008 Chr.12: 6977727-6977727 on Build GRCh38
rs782802160 Chr.12: 6977705-6977705 on Build GRCh38
rs782703712 Chr.12: 6977674-6977674 on Build GRCh38
rs781872400 Chr.12: 6977689-6977689 on Build GRCh38
rs782081649 Chr.12: 6977664-6977664 on Build GRCh38
rs781863798 Chr.12: 6977719-6977719 on Build GRCh38
rs7488179 Chr.12: 6977654-6977654 on Build GRCh38
rs781793429 Chr.12: 6977769-6977769 on Build GRCh38
rs80101148 Chr.12: 6977740-6977740 on Build GRCh38
rs782597754 Chr.12: 6977585-6977585 on Build GRCh38
rs782016708 Chr.12: 6977600-6977600 on Build GRCh38
rs782264457 Chr.12: 6977607-6977607 on Build GRCh38
rs140765444 Chr.12: 6977688-6977688 on Build GRCh38
rs782468847 Chr.12: 6977579-6977579 on Build GRCh38
rs782514016 Chr.12: 6977768-6977768 on Build GRCh38
rs372817408 Chr.12: 6977594-6977594 on Build GRCh38
rs782178630 Chr.12: 6977586-6977586 on Build GRCh38
rs781891178 Chr.12: 6977763-6977763 on Build GRCh38
rs772868236 Chr.12: 6977603-6977603 on Build GRCh38
rs782100333 Chr.12: 6977617-6977617 on Build GRCh38
rs781965255 Chr.12: 6977637-6977637 on Build GRCh38

View Full Product Details