Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 50961781-50962034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLK6
Entrez Gene ID
5653
Gene Name
kallikrein related peptidase 6
Gene Aliases
Bssp, Klk7, PRSS18, PRSS9, SP59, hK6
UniGene
Hs.79361
Gene Chromosome Location
Chr.19: 50958631-50969673 on Build GRCh38
Transcripts
NM_001012964.2 NM_001012965.2 NM_001319948.1 NM_001319949.1 NM_002774.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6476946 c.536A>C p.N179T Chr.19: 50961790-50961790 on Build GRCh38
4894901 c.449A>G p.D150G Chr.19: 50961877-50961877 on Build GRCh38
3537423 c.521G>A p.G174D Chr.19: 50961805-50961805 on Build GRCh38
385047 c.472G>C p.A158P Chr.19: 50961854-50961854 on Build GRCh38
3537424 c.454C>T p.P152S Chr.19: 50961872-50961872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775873008 Chr.19: 50961807-50961807 on Build GRCh38
rs376502607 Chr.19: 50961920-50961920 on Build GRCh38
rs1021139548 Chr.19: 50961976-50961976 on Build GRCh38
rs752199342 Chr.19: 50961821-50961821 on Build GRCh38
rs988437413 Chr.19: 50961954-50961954 on Build GRCh38
rs561945287 Chr.19: 50961980-50961980 on Build GRCh38
rs779738082 Chr.19: 50961861-50961861 on Build GRCh38
rs1010540712 Chr.19: 50961949-50961949 on Build GRCh38
rs761097805 Chr.19: 50961910-50961910 on Build GRCh38
rs927772869 Chr.19: 50962002-50962002 on Build GRCh38
rs571510641 Chr.19: 50961909-50961909 on Build GRCh38
rs762495499 Chr.19: 50961826-50961826 on Build GRCh38
rs775756729 Chr.19: 50961907-50961907 on Build GRCh38
rs976433834 Chr.19: 50961981-50961981 on Build GRCh38
rs748036646 Chr.19: 50961890-50961890 on Build GRCh38
rs769430408 Chr.19: 50961892-50961892 on Build GRCh38
rs368882893 Chr.19: 50961843-50961843 on Build GRCh38
rs772333292 Chr.19: 50961953-50961953 on Build GRCh38
rs145562169 Chr.19: 50961919-50961919 on Build GRCh38
rs1032755698 Chr.19: 50961936-50961936 on Build GRCh38
rs538837553 Chr.19: 50961912-50961912 on Build GRCh38
rs921785614 Chr.19: 50961982-50961982 on Build GRCh38
rs1040509198 Chr.19: 50961818-50961818 on Build GRCh38
rs957949162 Chr.19: 50961933-50961933 on Build GRCh38
rs1020134327 Chr.19: 50961963-50961963 on Build GRCh38
rs748970718 Chr.19: 50961893-50961893 on Build GRCh38
rs761134835 Chr.19: 50961809-50961809 on Build GRCh38
rs768388862 Chr.19: 50961832-50961832 on Build GRCh38
rs144453580 Chr.19: 50961883-50961883 on Build GRCh38
rs1014144202 Chr.19: 50961852-50961852 on Build GRCh38
rs957000088 Chr.19: 50961942-50961942 on Build GRCh38
rs140476582 Chr.19: 50961842-50961842 on Build GRCh38
rs751349589 Chr.19: 50961865-50961865 on Build GRCh38
rs931842708 Chr.19: 50961984-50961984 on Build GRCh38
rs981970206 Chr.19: 50961993-50961993 on Build GRCh38
rs770683890 Chr.19: 50961904-50961904 on Build GRCh38
rs1701950 Chr.19: 50961840-50961840 on Build GRCh38
rs112376245 Chr.19: 50961944-50961945 on Build GRCh38
rs201461622 Chr.19: 50961872-50961872 on Build GRCh38
rs754934254 Chr.19: 50961880-50961880 on Build GRCh38
rs765335845 Chr.19: 50961926-50961926 on Build GRCh38
rs764632929 Chr.19: 50961817-50961817 on Build GRCh38
rs984860421 Chr.19: 50962001-50962001 on Build GRCh38
rs996384137 Chr.19: 50961845-50961845 on Build GRCh38

View Full Product Details