Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 26919435-26919662 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLAA
Entrez Gene ID
9373
Gene Name
phospholipase A2 activating protein
Gene Aliases
DOA1, PLA2P, PLAP
UniGene
Hs.27182
Gene Chromosome Location
Chr.9: 26903370-26947473 on Build GRCh38
Transcripts
NM_001031689.2 NM_001321546.1 XM_011518072.2 XM_017015319.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4809410 c.1204G>C p.D402H Chr.9: 26919523-26919523 on Build GRCh38
3656690 c.1041T>A p.V347V Chr.9: 26919515-26919515 on Build GRCh38
4892095 c.1212T>A p.V404V Chr.9: 26919515-26919515 on Build GRCh38
3656689 c.1069C>G p.P357A Chr.9: 26919487-26919487 on Build GRCh38
4899207 c.1240C>G p.P414A Chr.9: 26919487-26919487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754862580 Chr.9: 26919576-26919576 on Build GRCh38
rs939009432 Chr.9: 26919471-26919471 on Build GRCh38
rs778431059 Chr.9: 26919473-26919473 on Build GRCh38
rs41271173 Chr.9: 26919595-26919595 on Build GRCh38
rs747241039 Chr.9: 26919613-26919616 on Build GRCh38
rs892585943 Chr.9: 26919624-26919624 on Build GRCh38
rs982033372 Chr.9: 26919607-26919607 on Build GRCh38
rs770146150 Chr.9: 26919519-26919519 on Build GRCh38
rs764232986 Chr.9: 26919538-26919538 on Build GRCh38
rs754593946 Chr.9: 26919467-26919467 on Build GRCh38
rs892171621 Chr.9: 26919524-26919524 on Build GRCh38
rs756156232 Chr.9: 26919555-26919555 on Build GRCh38
rs199972754 Chr.9: 26919504-26919504 on Build GRCh38
rs746423070 Chr.9: 26919508-26919508 on Build GRCh38
rs10120313 Chr.9: 26919542-26919542 on Build GRCh38
rs748934424 Chr.9: 26919463-26919463 on Build GRCh38
rs200655405 Chr.9: 26919461-26919461 on Build GRCh38
rs775748434 Chr.9: 26919527-26919527 on Build GRCh38
rs766133890 Chr.9: 26919566-26919566 on Build GRCh38
rs761427082 Chr.9: 26919559-26919559 on Build GRCh38
rs1050374131 Chr.9: 26919582-26919582 on Build GRCh38
rs763167708 Chr.9: 26919529-26919529 on Build GRCh38
rs767742217 Chr.9: 26919550-26919550 on Build GRCh38
rs761977522 Chr.9: 26919545-26919545 on Build GRCh38
rs376987716 Chr.9: 26919485-26919485 on Build GRCh38
rs750360408 Chr.9: 26919551-26919551 on Build GRCh38
rs890428425 Chr.9: 26919586-26919586 on Build GRCh38
rs10120342 Chr.9: 26919609-26919609 on Build GRCh38
rs753591699 Chr.9: 26919575-26919575 on Build GRCh38
rs974387357 Chr.9: 26919635-26919635 on Build GRCh38
rs142993896 Chr.9: 26919627-26919627 on Build GRCh38
rs747645310 Chr.9: 26919475-26919475 on Build GRCh38

View Full Product Details