Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10392882-10393131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH8
Entrez Gene ID
4626
Gene Name
myosin heavy chain 8
Gene Aliases
DA7, MyHC-peri, MyHC-pn, gtMHC-F
UniGene
Hs.700484
Gene Chromosome Location
Chr.17: 10390325-10421950 on Build GRCh38
Transcripts
NM_002472.2 NR_125367.1

Gene Symbol
MYHAS
Entrez Gene ID
100128560
Gene Name
myosin heavy chain gene cluster antisense RNA
Gene Aliases
-
UniGene
Hs.661174
Gene Chromosome Location
Chr.17: 10383132-10625540 on Build GRCh38
Transcripts
NM_002472.2 NR_125367.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3513626 c.5408C>T p.A1803V Chr.17: 10392886-10392886 on Build GRCh38
3513627 c.5282C>T p.A1761V Chr.17: 10393095-10393095 on Build GRCh38
3733901 c.5375C>T p.T1792M Chr.17: 10392919-10392919 on Build GRCh38
559519 c.5395C>T p.R1799C Chr.17: 10392899-10392899 on Build GRCh38
325894 c.5406G>T p.E1802D Chr.17: 10392888-10392888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748681248 Chr.17: 10392931-10392933 on Build GRCh38
rs759676157 Chr.17: 10393083-10393083 on Build GRCh38
rs774984611 Chr.17: 10393038-10393038 on Build GRCh38
rs764923155 Chr.17: 10393065-10393065 on Build GRCh38
rs767783112 Chr.17: 10393085-10393085 on Build GRCh38
rs892572649 Chr.17: 10393040-10393040 on Build GRCh38
rs756213751 Chr.17: 10393089-10393089 on Build GRCh38
rs577510735 Chr.17: 10392964-10392964 on Build GRCh38
rs768599210 Chr.17: 10393056-10393056 on Build GRCh38
rs563469677 Chr.17: 10393096-10393096 on Build GRCh38
rs541994027 Chr.17: 10393066-10393066 on Build GRCh38
rs200951146 Chr.17: 10392918-10392918 on Build GRCh38
rs770630534 Chr.17: 10392908-10392908 on Build GRCh38
rs267604699 Chr.17: 10392990-10392990 on Build GRCh38
rs1010708924 Chr.17: 10393037-10393037 on Build GRCh38
rs367712075 Chr.17: 10392969-10392969 on Build GRCh38
rs753640473 Chr.17: 10392945-10392945 on Build GRCh38
rs773323805 Chr.17: 10393103-10393104 on Build GRCh38
rs775461673 Chr.17: 10392942-10392942 on Build GRCh38
rs776508765 Chr.17: 10393057-10393057 on Build GRCh38
rs529647820 Chr.17: 10392957-10392957 on Build GRCh38
rs769827979 Chr.17: 10393073-10393074 on Build GRCh38
rs771654542 Chr.17: 10392998-10392998 on Build GRCh38
rs752840291 Chr.17: 10393088-10393088 on Build GRCh38
rs141215006 Chr.17: 10392944-10392944 on Build GRCh38
rs1053856177 Chr.17: 10393017-10393017 on Build GRCh38
rs745493875 Chr.17: 10393018-10393018 on Build GRCh38
rs138165573 Chr.17: 10392943-10392943 on Build GRCh38
rs200906285 Chr.17: 10392956-10392956 on Build GRCh38
rs371184733 Chr.17: 10392976-10392976 on Build GRCh38
rs778675991 Chr.17: 10393004-10393004 on Build GRCh38
rs199803469 Chr.17: 10392983-10392983 on Build GRCh38
rs777179030 Chr.17: 10393059-10393059 on Build GRCh38
rs369896488 Chr.17: 10392936-10392936 on Build GRCh38
rs760117922 Chr.17: 10393043-10393043 on Build GRCh38
rs144785726 Chr.17: 10393002-10393002 on Build GRCh38
rs267604698 Chr.17: 10392912-10392912 on Build GRCh38
rs144962215 Chr.17: 10392919-10392919 on Build GRCh38
rs574711257 Chr.17: 10393051-10393051 on Build GRCh38
rs751704006 Chr.17: 10392959-10392959 on Build GRCh38
rs753326378 Chr.17: 10392971-10392971 on Build GRCh38
rs749692404 Chr.17: 10393001-10393001 on Build GRCh38
rs771677133 Chr.17: 10393027-10393027 on Build GRCh38
rs781121685 Chr.17: 10392966-10392966 on Build GRCh38

View Full Product Details