Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75899441-75899714 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRPL38
Entrez Gene ID
64978
Gene Name
mitochondrial ribosomal protein L38
Gene Aliases
HSPC262, L38MT, MRP-L3, MRP-L38, RPML3
UniGene
Hs.442609
Gene Chromosome Location
Chr.17: 75898643-75905100 on Build GRCh38
Transcripts
NM_032478.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1386075 c.731G>A p.R244Q Chr.17: 75899654-75899654 on Build GRCh38
4069720 c.864A>G p.S288S Chr.17: 75899521-75899521 on Build GRCh38
1386074 c.771_772insC p.F258fs*23 Chr.17: 75899613-75899614 on Build GRCh38
4703217 c.771delC p.F258fs*54 Chr.17: 75899614-75899614 on Build GRCh38
6279856 c.736G>A p.A246T Chr.17: 75899649-75899649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779303444 Chr.17: 75899656-75899656 on Build GRCh38
rs1015315397 Chr.17: 75899635-75899635 on Build GRCh38
rs200298025 Chr.17: 75899532-75899532 on Build GRCh38
rs776372050 Chr.17: 75899685-75899685 on Build GRCh38
rs763318116 Chr.17: 75899519-75899519 on Build GRCh38
rs566410319 Chr.17: 75899529-75899529 on Build GRCh38
rs772425113 Chr.17: 75899616-75899616 on Build GRCh38
rs765235241 Chr.17: 75899596-75899596 on Build GRCh38
rs148417989 Chr.17: 75899662-75899662 on Build GRCh38
rs1128878 Chr.17: 75899663-75899663 on Build GRCh38
rs753952457 Chr.17: 75899655-75899655 on Build GRCh38
rs199719387 Chr.17: 75899583-75899583 on Build GRCh38
rs552586623 Chr.17: 75899488-75899488 on Build GRCh38
rs757308577 Chr.17: 75899595-75899595 on Build GRCh38
rs370453564 Chr.17: 75899505-75899505 on Build GRCh38
rs767075035 Chr.17: 75899542-75899542 on Build GRCh38
rs766658607 Chr.17: 75899525-75899525 on Build GRCh38
rs760404060 Chr.17: 75899652-75899652 on Build GRCh38
rs368094656 Chr.17: 75899614-75899614 on Build GRCh38
rs752242629 Chr.17: 75899471-75899471 on Build GRCh38
rs769805492 Chr.17: 75899558-75899558 on Build GRCh38
rs759329353 Chr.17: 75899497-75899497 on Build GRCh38
rs375469013 Chr.17: 75899628-75899628 on Build GRCh38
rs372172749 Chr.17: 75899691-75899691 on Build GRCh38
rs768908572 Chr.17: 75899496-75899496 on Build GRCh38
rs756056986 Chr.17: 75899480-75899480 on Build GRCh38
rs768454084 Chr.17: 75899681-75899681 on Build GRCh38
rs191961404 Chr.17: 75899575-75899575 on Build GRCh38
rs777570823 Chr.17: 75899493-75899494 on Build GRCh38
rs761366700 Chr.17: 75899622-75899622 on Build GRCh38
rs532169101 Chr.17: 75899482-75899482 on Build GRCh38
rs763630440 Chr.17: 75899586-75899586 on Build GRCh38
rs758439223 Chr.17: 75899614-75899614 on Build GRCh38
rs775012277 Chr.17: 75899617-75899617 on Build GRCh38
rs375231046 Chr.17: 75899557-75899557 on Build GRCh38
rs767259396 Chr.17: 75899613-75899613 on Build GRCh38
rs766259000 Chr.17: 75899633-75899633 on Build GRCh38
rs140320185 Chr.17: 75899604-75899604 on Build GRCh38
rs55843905 Chr.17: 75899607-75899607 on Build GRCh38
rs761919516 Chr.17: 75899500-75899500 on Build GRCh38
rs563116995 Chr.17: 75899464-75899464 on Build GRCh38
rs746322017 Chr.17: 75899660-75899660 on Build GRCh38
rs372305971 Chr.17: 75899615-75899615 on Build GRCh38
rs948288510 Chr.17: 75899463-75899463 on Build GRCh38
rs562216718 Chr.17: 75899545-75899545 on Build GRCh38
rs747345745 Chr.17: 75899665-75899665 on Build GRCh38
rs150890281 Chr.17: 75899540-75899540 on Build GRCh38
rs780016287 Chr.17: 75899613-75899614 on Build GRCh38
rs143444007 Chr.17: 75899551-75899551 on Build GRCh38
rs928223193 Chr.17: 75899689-75899689 on Build GRCh38
rs781434507 Chr.17: 75899552-75899552 on Build GRCh38
rs368523779 Chr.17: 75899645-75899645 on Build GRCh38
rs780436781 Chr.17: 75899489-75899489 on Build GRCh38
rs866385646 Chr.17: 75899612-75899613 on Build GRCh38
rs773708468 Chr.17: 75899559-75899559 on Build GRCh38
rs773657184 Chr.17: 75899518-75899518 on Build GRCh38
rs572018825 Chr.17: 75899620-75899620 on Build GRCh38
rs753394159 Chr.17: 75899587-75899587 on Build GRCh38
rs749127275 Chr.17: 75899484-75899484 on Build GRCh38
rs140072876 Chr.17: 75899548-75899548 on Build GRCh38
rs145698092 Chr.17: 75899568-75899568 on Build GRCh38
rs746915005 Chr.17: 75899618-75899619 on Build GRCh38
rs540793162 Chr.17: 75899632-75899632 on Build GRCh38
rs147988641 Chr.17: 75899571-75899571 on Build GRCh38
rs758130664 Chr.17: 75899631-75899631 on Build GRCh38
rs147320099 Chr.17: 75899603-75899603 on Build GRCh38
rs749856620 Chr.17: 75899566-75899566 on Build GRCh38
rs767109741 Chr.17: 75899578-75899578 on Build GRCh38
rs759172796 Chr.17: 75899528-75899528 on Build GRCh38
rs751913463 Chr.17: 75899526-75899526 on Build GRCh38
rs145518548 Chr.17: 75899654-75899654 on Build GRCh38
rs777876516 Chr.17: 75899533-75899533 on Build GRCh38
rs765286447 Chr.17: 75899626-75899626 on Build GRCh38
rs760470974 Chr.17: 75899618-75899618 on Build GRCh38
rs760117849 Chr.17: 75899585-75899585 on Build GRCh38
rs752191184 Chr.17: 75899531-75899531 on Build GRCh38

View Full Product Details