Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 58874197-58874424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRIG3
Entrez Gene ID
121227
Gene Name
leucine rich repeats and immunoglobulin like domains 3
Gene Aliases
LIG3
UniGene
Hs.253736
Gene Chromosome Location
Chr.12: 58869351-58920540 on Build GRCh38
Transcripts
NM_001136051.2 NM_153377.4 XM_011537893.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5172473 c.2660-2_2660-1insA p.? Chr.12: 58874331-58874332 on Build GRCh38
5601752 c.2786C>T p.S929L Chr.12: 58874204-58874204 on Build GRCh38
5909252 c.2840G>A p.G947D Chr.12: 58874330-58874330 on Build GRCh38
942269 c.2944G>A p.E982K Chr.12: 58874226-58874226 on Build GRCh38
942270 c.2841T>C p.G947G Chr.12: 58874329-58874329 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142618940 Chr.12: 58874349-58874350 on Build GRCh38
rs761355712 Chr.12: 58874313-58874313 on Build GRCh38
rs950643019 Chr.12: 58874238-58874238 on Build GRCh38
rs760327875 Chr.12: 58874347-58874348 on Build GRCh38
rs757083237 Chr.12: 58874237-58874237 on Build GRCh38
rs760209970 Chr.12: 58874300-58874300 on Build GRCh38
rs1057031337 Chr.12: 58874367-58874367 on Build GRCh38
rs768811408 Chr.12: 58874254-58874254 on Build GRCh38
rs750185700 Chr.12: 58874339-58874339 on Build GRCh38
rs773482598 Chr.12: 58874222-58874223 on Build GRCh38
rs771286510 Chr.12: 58874394-58874394 on Build GRCh38
rs753738730 Chr.12: 58874222-58874222 on Build GRCh38
rs774878508 Chr.12: 58874399-58874399 on Build GRCh38
rs140112794 Chr.12: 58874227-58874227 on Build GRCh38
rs769905460 Chr.12: 58874373-58874373 on Build GRCh38
rs146864816 Chr.12: 58874315-58874315 on Build GRCh38
rs778735593 Chr.12: 58874385-58874388 on Build GRCh38
rs202041991 Chr.12: 58874223-58874223 on Build GRCh38
rs763702006 Chr.12: 58874350-58874354 on Build GRCh38
rs749629687 Chr.12: 58874377-58874377 on Build GRCh38
rs10877180 Chr.12: 58874347-58874347 on Build GRCh38
rs759235769 Chr.12: 58874278-58874279 on Build GRCh38
rs777985306 Chr.12: 58874375-58874375 on Build GRCh38
rs143823251 Chr.12: 58874259-58874259 on Build GRCh38
rs751505955 Chr.12: 58874344-58874344 on Build GRCh38
rs764421199 Chr.12: 58874354-58874356 on Build GRCh38
rs747102590 Chr.12: 58874252-58874252 on Build GRCh38
rs779881279 Chr.12: 58874343-58874343 on Build GRCh38
rs773437574 Chr.12: 58874283-58874283 on Build GRCh38
rs763584366 Chr.12: 58874289-58874289 on Build GRCh38
rs963536342 Chr.12: 58874288-58874288 on Build GRCh38
rs748441187 Chr.12: 58874363-58874363 on Build GRCh38
rs752245736 Chr.12: 58874341-58874342 on Build GRCh38
rs779972316 Chr.12: 58874247-58874247 on Build GRCh38
rs753535719 Chr.12: 58874303-58874303 on Build GRCh38
rs546069057 Chr.12: 58874340-58874340 on Build GRCh38
rs201291215 Chr.12: 58874361-58874361 on Build GRCh38
rs1030268817 Chr.12: 58874228-58874228 on Build GRCh38
rs767331385 Chr.12: 58874326-58874327 on Build GRCh38
rs373257232 Chr.12: 58874270-58874270 on Build GRCh38
rs954036491 Chr.12: 58874226-58874226 on Build GRCh38
rs151124358 Chr.12: 58874336-58874336 on Build GRCh38
rs753447814 Chr.12: 58874235-58874235 on Build GRCh38
rs750430621 Chr.12: 58874242-58874242 on Build GRCh38
rs763562669 Chr.12: 58874301-58874301 on Build GRCh38
rs377330807 Chr.12: 58874277-58874277 on Build GRCh38
rs200278478 Chr.12: 58874359-58874359 on Build GRCh38
rs563804582 Chr.12: 58874244-58874244 on Build GRCh38
rs937644751 Chr.12: 58874379-58874379 on Build GRCh38
rs369103183 Chr.12: 58874240-58874240 on Build GRCh38
rs770144147 Chr.12: 58874221-58874222 on Build GRCh38
rs771347438 Chr.12: 58874293-58874293 on Build GRCh38
rs774790509 Chr.12: 58874295-58874295 on Build GRCh38
rs113449024 Chr.12: 58874249-58874249 on Build GRCh38
rs757166500 Chr.12: 58874356-58874357 on Build GRCh38

View Full Product Details