Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 200173956-200174219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NR5A2
Entrez Gene ID
2494
Gene Name
nuclear receptor subfamily 5 group A member 2
Gene Aliases
B1F, B1F2, CPF, FTF, FTZ-F1, FTZ-F1beta, LRH-1, LRH1, hB1F-2
UniGene
Hs.33446
Gene Chromosome Location
Chr.1: 200027602-200177424 on Build GRCh38
Transcripts
NM_001276464.1 NM_003822.4 NM_205860.2 XM_005245062.3 XM_011509380.1 XM_011509381.2 XM_011509382.1 XM_011509384.2 XM_017000904.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
363171 c.1403A>T p.Q468L Chr.1: 200173987-200173987 on Build GRCh38
5496437 c.1484A>G p.K495R Chr.1: 200174068-200174068 on Build GRCh38
4423877 c.1379delA p.D460fs*11 Chr.1: 200173963-200173963 on Build GRCh38
3664967 c.1428A>G p.Q476Q Chr.1: 200174012-200174012 on Build GRCh38
5497585 c.1470G>T p.P490P Chr.1: 200174054-200174054 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs4255412 Chr.1: 200174127-200174127 on Build GRCh38
rs764311097 Chr.1: 200174156-200174156 on Build GRCh38
rs777167598 Chr.1: 200174115-200174115 on Build GRCh38
rs1015956920 Chr.1: 200174147-200174147 on Build GRCh38
rs778784980 Chr.1: 200174187-200174187 on Build GRCh38
rs1050922837 Chr.1: 200174097-200174097 on Build GRCh38
rs765564537 Chr.1: 200174017-200174017 on Build GRCh38
rs183953069 Chr.1: 200174053-200174053 on Build GRCh38
rs139798381 Chr.1: 200174180-200174180 on Build GRCh38
rs757388795 Chr.1: 200174165-200174165 on Build GRCh38
rs751586641 Chr.1: 200174157-200174157 on Build GRCh38
rs147419125 Chr.1: 200174099-200174099 on Build GRCh38
rs771306384 Chr.1: 200174104-200174104 on Build GRCh38
rs766924985 Chr.1: 200173988-200173988 on Build GRCh38
rs768466796 Chr.1: 200174066-200174066 on Build GRCh38
rs747092960 Chr.1: 200174022-200174022 on Build GRCh38
rs202153830 Chr.1: 200174003-200174003 on Build GRCh38
rs748994632 Chr.1: 200174054-200174054 on Build GRCh38
rs946976621 Chr.1: 200174071-200174071 on Build GRCh38
rs143039367 Chr.1: 200174087-200174087 on Build GRCh38
rs760015255 Chr.1: 200174120-200174120 on Build GRCh38
rs995931179 Chr.1: 200174154-200174154 on Build GRCh38
rs150243341 Chr.1: 200174021-200174021 on Build GRCh38
rs373453960 Chr.1: 200174152-200174152 on Build GRCh38
rs771855360 Chr.1: 200174096-200174096 on Build GRCh38
rs565841747 Chr.1: 200174102-200174102 on Build GRCh38
rs149054125 Chr.1: 200174048-200174048 on Build GRCh38
rs752897392 Chr.1: 200174020-200174020 on Build GRCh38
rs747798997 Chr.1: 200174089-200174089 on Build GRCh38
rs778425009 Chr.1: 200174020-200174020 on Build GRCh38
rs376108677 Chr.1: 200174182-200174182 on Build GRCh38
rs932350625 Chr.1: 200174042-200174042 on Build GRCh38
rs1049925447 Chr.1: 200174036-200174036 on Build GRCh38
rs781167513 Chr.1: 200174166-200174166 on Build GRCh38
rs754105342 Chr.1: 200174000-200174000 on Build GRCh38
rs765625573 Chr.1: 200174132-200174132 on Build GRCh38
rs1060060 Chr.1: 200174153-200174153 on Build GRCh38

View Full Product Details