Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5207903-5208128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPRS
Entrez Gene ID
5802
Gene Name
protein tyrosine phosphatase, receptor type S
Gene Aliases
PTPSIGMA, R-PTP-S, R-PTP-sigma
UniGene
Hs.744928
Gene Chromosome Location
Chr.19: 5205503-5340803 on Build GRCh38
Transcripts
NM_002850.3 NM_130853.2 NM_130854.2 NM_130855.2 XM_005259600.2 XM_005259606.2 XM_005259607.2 XM_005259609.1 XM_005259610.1 XM_011528157.2 XM_011528158.2 XM_017027065.1 XM_017027066.1 XM_017027067.1 XM_017027068.1 XM_017027069.1 XM_017027070.1 XM_017027071.1 XM_017027072.1 XM_017027073.1 XM_017027074.1 XM_017027075.1 XM_017027076.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6960817 c.5694G>T p.E1898D Chr.19: 5208006-5208006 on Build GRCh38
4783049 c.5699G>A p.R1900Q Chr.19: 5208004-5208004 on Build GRCh38
4904105 c.5688C>T p.I1896I Chr.19: 5208015-5208015 on Build GRCh38
276959 c.5660C>T p.T1887M Chr.19: 5208040-5208040 on Build GRCh38
6915724 c.5701C>T p.R1901W Chr.19: 5207999-5207999 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772956659 Chr.19: 5208025-5208025 on Build GRCh38
rs139519201 Chr.19: 5208047-5208047 on Build GRCh38
rs767854985 Chr.19: 5208079-5208079 on Build GRCh38
rs753240756 Chr.19: 5207964-5207964 on Build GRCh38
rs769436280 Chr.19: 5208105-5208105 on Build GRCh38
rs35508296 Chr.19: 5208006-5208006 on Build GRCh38
rs774101179 Chr.19: 5208039-5208039 on Build GRCh38
rs369125705 Chr.19: 5208104-5208104 on Build GRCh38
rs375237464 Chr.19: 5207928-5207928 on Build GRCh38
rs778830949 Chr.19: 5208090-5208090 on Build GRCh38
rs767844549 Chr.19: 5207959-5207959 on Build GRCh38
rs372859087 Chr.19: 5208024-5208024 on Build GRCh38
rs752778384 Chr.19: 5208089-5208089 on Build GRCh38
rs762312580 Chr.19: 5207936-5207936 on Build GRCh38
rs968335743 Chr.19: 5207941-5207941 on Build GRCh38
rs749395911 Chr.19: 5207980-5207980 on Build GRCh38
rs758182581 Chr.19: 5208089-5208100 on Build GRCh38
rs115470613 Chr.19: 5207968-5207968 on Build GRCh38
rs145958658 Chr.19: 5208041-5208041 on Build GRCh38
rs942270803 Chr.19: 5208017-5208017 on Build GRCh38
rs757213585 Chr.19: 5207987-5207987 on Build GRCh38
rs138437349 Chr.19: 5208015-5208015 on Build GRCh38
rs917980770 Chr.19: 5207970-5207970 on Build GRCh38
rs145778823 Chr.19: 5207967-5207967 on Build GRCh38
rs759155627 Chr.19: 5208050-5208050 on Build GRCh38
rs141597386 Chr.19: 5207949-5207949 on Build GRCh38
rs752031088 Chr.19: 5208004-5208004 on Build GRCh38
rs200796184 Chr.19: 5208054-5208054 on Build GRCh38
rs768615091 Chr.19: 5208014-5208014 on Build GRCh38
rs755356872 Chr.19: 5207958-5207958 on Build GRCh38
rs549497966 Chr.19: 5208075-5208075 on Build GRCh38
rs142977438 Chr.19: 5208035-5208035 on Build GRCh38
rs753044393 Chr.19: 5208053-5208053 on Build GRCh38
rs758681871 Chr.19: 5208101-5208101 on Build GRCh38
rs766060117 Chr.19: 5207940-5207940 on Build GRCh38
rs571188959 Chr.19: 5208092-5208092 on Build GRCh38
rs757411260 Chr.19: 5208087-5208087 on Build GRCh38
rs1042776388 Chr.19: 5208005-5208005 on Build GRCh38
rs531465256 Chr.19: 5208045-5208045 on Build GRCh38
rs368528400 Chr.19: 5207988-5207988 on Build GRCh38
rs746059446 Chr.19: 5207998-5207998 on Build GRCh38
rs755196138 Chr.19: 5208036-5208036 on Build GRCh38
rs748156971 Chr.19: 5208022-5208022 on Build GRCh38
rs764663502 Chr.19: 5208074-5208074 on Build GRCh38
rs780367081 Chr.19: 5208103-5208103 on Build GRCh38
rs767934428 Chr.19: 5208051-5208051 on Build GRCh38

View Full Product Details