Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 155083983-155084254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MME
Entrez Gene ID
4311
Gene Name
membrane metallo-endopeptidase
Gene Aliases
CALLA, CD10, CMT2T, NEP, SFE
UniGene
Hs.307734
Gene Chromosome Location
Chr.3: 155024124-155183729 on Build GRCh38
Transcripts
NM_000902.3 NM_007287.2 NM_007288.2 NM_007289.2 XM_006713646.3 XM_006713647.3 XM_011512855.2 XM_011512856.2 XM_011512857.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6544963 c.84G>T p.E28D Chr.3: 155084251-155084251 on Build GRCh38
6096768 c.87_88CA>AG p.S30G Chr.3: 155084254-155084255 on Build GRCh38
3588992 c.82G>A p.E28K Chr.3: 155084249-155084249 on Build GRCh38
1040241 c.67C>T p.R23* Chr.3: 155084234-155084234 on Build GRCh38
3588991 c.76C>T p.P26S Chr.3: 155084243-155084243 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534436536 Chr.3: 155084087-155084087 on Build GRCh38
rs551664278 Chr.3: 155084055-155084055 on Build GRCh38
rs61762318 Chr.3: 155084135-155084135 on Build GRCh38
rs755684422 Chr.3: 155084220-155084220 on Build GRCh38
rs775680504 Chr.3: 155084185-155084185 on Build GRCh38
rs61762319 Chr.3: 155084189-155084189 on Build GRCh38
rs150836510 Chr.3: 155084234-155084234 on Build GRCh38
rs766989448 Chr.3: 155084212-155084212 on Build GRCh38
rs978354507 Chr.3: 155084219-155084219 on Build GRCh38
rs56208271 Chr.3: 155084043-155084043 on Build GRCh38
rs866991983 Chr.3: 155084224-155084224 on Build GRCh38
rs148774018 Chr.3: 155084141-155084141 on Build GRCh38
rs934068733 Chr.3: 155084026-155084026 on Build GRCh38
rs772274287 Chr.3: 155084179-155084179 on Build GRCh38
rs199935778 Chr.3: 155084214-155084214 on Build GRCh38
rs764401706 Chr.3: 155084190-155084190 on Build GRCh38
rs1051147733 Chr.3: 155084029-155084029 on Build GRCh38
rs751490017 Chr.3: 155084205-155084205 on Build GRCh38
rs774811801 Chr.3: 155084147-155084148 on Build GRCh38
rs747720840 Chr.3: 155084123-155084123 on Build GRCh38
rs550863574 Chr.3: 155084035-155084035 on Build GRCh38
rs141402067 Chr.3: 155084233-155084233 on Build GRCh38
rs757011311 Chr.3: 155084134-155084134 on Build GRCh38
rs914931190 Chr.3: 155084207-155084207 on Build GRCh38
rs28673360 Chr.3: 155084013-155084013 on Build GRCh38
rs755224674 Chr.3: 155084089-155084089 on Build GRCh38
rs924198803 Chr.3: 155084221-155084221 on Build GRCh38
rs762748390 Chr.3: 155084193-155084193 on Build GRCh38
rs753755102 Chr.3: 155084115-155084115 on Build GRCh38
rs868086674 Chr.3: 155084049-155084049 on Build GRCh38
rs766220341 Chr.3: 155084204-155084204 on Build GRCh38
rs148334392 Chr.3: 155084223-155084223 on Build GRCh38
rs902724736 Chr.3: 155084022-155084022 on Build GRCh38
rs569202340 Chr.3: 155084036-155084036 on Build GRCh38
rs199848907 Chr.3: 155084159-155084159 on Build GRCh38
rs1012149293 Chr.3: 155084137-155084137 on Build GRCh38
rs746439998 Chr.3: 155084178-155084178 on Build GRCh38
rs567029551 Chr.3: 155084074-155084074 on Build GRCh38
rs200493354 Chr.3: 155084166-155084166 on Build GRCh38
rs777597280 Chr.3: 155084095-155084095 on Build GRCh38
rs767186383 Chr.3: 155084020-155084020 on Build GRCh38
rs142421146 Chr.3: 155084209-155084209 on Build GRCh38
rs1034739692 Chr.3: 155084156-155084156 on Build GRCh38
rs370911005 Chr.3: 155084192-155084192 on Build GRCh38

View Full Product Details