Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41457735-41457960 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTF1
Entrez Gene ID
23168
Gene Name
RTF1 homolog, Paf1/RNA polymerase II complex component
Gene Aliases
GTL7, KIAA0252
UniGene
Hs.511096
Gene Chromosome Location
Chr.15: 41417104-41483563 on Build GRCh38
Transcripts
NM_015138.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849048 c.226G>A p.E76K Chr.15: 41457815-41457815 on Build GRCh38
4849041 c.279G>C p.L93F Chr.15: 41457868-41457868 on Build GRCh38
4849020 c.227A>T p.E76V Chr.15: 41457816-41457816 on Build GRCh38
6270127 c.193G>T p.E65* Chr.15: 41457782-41457782 on Build GRCh38
7152169 c.193G>A p.E65K Chr.15: 41457782-41457782 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751360176 Chr.15: 41457807-41457807 on Build GRCh38
rs746399909 Chr.15: 41457927-41457927 on Build GRCh38
rs899614751 Chr.15: 41457840-41457840 on Build GRCh38
rs763605898 Chr.15: 41457928-41457929 on Build GRCh38
rs143376639 Chr.15: 41457814-41457814 on Build GRCh38
rs759880693 Chr.15: 41457899-41457899 on Build GRCh38
rs796779786 Chr.15: 41457899-41457900 on Build GRCh38
rs748411129 Chr.15: 41457841-41457841 on Build GRCh38
rs137930710 Chr.15: 41457808-41457808 on Build GRCh38
rs777856365 Chr.15: 41457871-41457871 on Build GRCh38
rs778790284 Chr.15: 41457921-41457921 on Build GRCh38
rs749423834 Chr.15: 41457914-41457914 on Build GRCh38
rs954640881 Chr.15: 41457856-41457856 on Build GRCh38
rs534694838 Chr.15: 41457766-41457766 on Build GRCh38
rs147835911 Chr.15: 41457823-41457823 on Build GRCh38
rs772489944 Chr.15: 41457901-41457901 on Build GRCh38
rs201885004 Chr.15: 41457905-41457905 on Build GRCh38
rs563644270 Chr.15: 41457907-41457907 on Build GRCh38
rs913073142 Chr.15: 41457896-41457896 on Build GRCh38
rs768784352 Chr.15: 41457868-41457869 on Build GRCh38
rs773582673 Chr.15: 41457906-41457907 on Build GRCh38
rs761579044 Chr.15: 41457903-41457903 on Build GRCh38
rs777946939 Chr.15: 41457909-41457909 on Build GRCh38
rs748467207 Chr.15: 41457898-41457899 on Build GRCh38
rs771473826 Chr.15: 41457884-41457884 on Build GRCh38
rs531654609 Chr.15: 41457899-41457900 on Build GRCh38
rs781056872 Chr.15: 41457908-41457908 on Build GRCh38
rs11313160 Chr.15: 41457899-41457899 on Build GRCh38
rs774903643 Chr.15: 41457893-41457893 on Build GRCh38
rs764944949 Chr.15: 41457904-41457904 on Build GRCh38
rs370585838 Chr.15: 41457784-41457784 on Build GRCh38
rs796683835 Chr.15: 41457907-41457907 on Build GRCh38
rs28645240 Chr.15: 41457821-41457821 on Build GRCh38
rs775658457 Chr.15: 41457902-41457902 on Build GRCh38
rs757350562 Chr.15: 41457916-41457916 on Build GRCh38
rs769948296 Chr.15: 41457869-41457869 on Build GRCh38
rs529781257 Chr.15: 41457879-41457879 on Build GRCh38

View Full Product Details