Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 86936782-86937013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MMRN2
Entrez Gene ID
79812
Gene Name
multimerin 2
Gene Aliases
EMILIN-3, EMILIN3, ENDOGLYX-1
UniGene
Hs.524479
Gene Chromosome Location
Chr.10: 86935540-86971311 on Build GRCh38
Transcripts
NM_024756.2 XM_005270153.1 XM_006717970.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6056228 c.2686C>T p.P896S Chr.10: 86936907-86936907 on Build GRCh38
7136180 c.2772_2773insT p.E925fs*1 Chr.10: 86936820-86936821 on Build GRCh38
6013971 c.2754G>C p.K918N Chr.10: 86936839-86936839 on Build GRCh38
4702747 c.2704C>T p.Q902* Chr.10: 86936889-86936889 on Build GRCh38
5999058 c.2675A>G p.H892R Chr.10: 86936918-86936918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753462877 Chr.10: 86936881-86936881 on Build GRCh38
rs760950750 Chr.10: 86936988-86936988 on Build GRCh38
rs756729371 Chr.10: 86936882-86936882 on Build GRCh38
rs1005494699 Chr.10: 86936937-86936937 on Build GRCh38
rs202191624 Chr.10: 86936993-86936993 on Build GRCh38
rs376535800 Chr.10: 86936852-86936852 on Build GRCh38
rs770988319 Chr.10: 86936860-86936860 on Build GRCh38
rs957946481 Chr.10: 86936871-86936871 on Build GRCh38
rs760433536 Chr.10: 86936876-86936876 on Build GRCh38
rs970525777 Chr.10: 86936854-86936854 on Build GRCh38
rs372692050 Chr.10: 86936833-86936833 on Build GRCh38
rs559930891 Chr.10: 86936923-86936923 on Build GRCh38
rs146646165 Chr.10: 86936855-86936855 on Build GRCh38
rs923339617 Chr.10: 86936859-86936859 on Build GRCh38
rs74794209 Chr.10: 86936837-86936837 on Build GRCh38
rs371020260 Chr.10: 86936913-86936913 on Build GRCh38
rs34587013 Chr.10: 86936865-86936865 on Build GRCh38
rs746783486 Chr.10: 86936905-86936905 on Build GRCh38
rs774863951 Chr.10: 86936863-86936863 on Build GRCh38
rs570128351 Chr.10: 86936867-86936867 on Build GRCh38
rs767910078 Chr.10: 86936866-86936866 on Build GRCh38
rs749927225 Chr.10: 86936994-86936994 on Build GRCh38
rs756270817 Chr.10: 86936954-86936954 on Build GRCh38
rs373201619 Chr.10: 86936926-86936926 on Build GRCh38
rs61740815 Chr.10: 86936941-86936941 on Build GRCh38
rs371372922 Chr.10: 86936967-86936967 on Build GRCh38
rs758377043 Chr.10: 86936893-86936893 on Build GRCh38
rs568385595 Chr.10: 86936848-86936848 on Build GRCh38
rs774384405 Chr.10: 86936947-86936947 on Build GRCh38
rs769580843 Chr.10: 86936868-86936868 on Build GRCh38
rs753274429 Chr.10: 86936808-86936808 on Build GRCh38
rs887041115 Chr.10: 86936932-86936932 on Build GRCh38
rs757284865 Chr.10: 86936811-86936811 on Build GRCh38
rs377460418 Chr.10: 86936966-86936966 on Build GRCh38
rs763649093 Chr.10: 86936877-86936877 on Build GRCh38
rs775899067 Chr.10: 86936985-86936985 on Build GRCh38
rs193180563 Chr.10: 86936989-86936989 on Build GRCh38
rs772012647 Chr.10: 86936832-86936832 on Build GRCh38
rs780039649 Chr.10: 86936900-86936900 on Build GRCh38
rs749191564 Chr.10: 86936981-86936984 on Build GRCh38
rs754739852 Chr.10: 86936909-86936909 on Build GRCh38
rs534261775 Chr.10: 86936868-86936869 on Build GRCh38
rs750415616 Chr.10: 86936887-86936887 on Build GRCh38
rs112224150 Chr.10: 86936818-86936818 on Build GRCh38
rs762950255 Chr.10: 86936856-86936856 on Build GRCh38
rs776167844 Chr.10: 86936990-86936990 on Build GRCh38
rs746090166 Chr.10: 86936826-86936826 on Build GRCh38
rs546562967 Chr.10: 86936831-86936831 on Build GRCh38

View Full Product Details