Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 69509023-69509252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OCLN
Entrez Gene ID
100506658
Gene Name
occludin
Gene Aliases
BLCPMG, PPP1R115
UniGene
Hs.592605
Gene Chromosome Location
Chr.5: 69492292-69558104 on Build GRCh38
Transcripts
NM_001205254.1 NM_001205255.1 NM_002538.3 XM_017008913.1 XM_017008914.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4790946 c.146A>G p.E49G Chr.5: 69509236-69509236 on Build GRCh38
7352536 c.70C>G p.P24A Chr.5: 69509160-69509160 on Build GRCh38
4775862 c.69A>G p.A23A Chr.5: 69509159-69509159 on Build GRCh38
4769738 c.143C>T p.P48L Chr.5: 69509233-69509233 on Build GRCh38
3661919 c.62A>G p.H21R Chr.5: 69509152-69509152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753732841 Chr.5: 69509211-69509211 on Build GRCh38
rs765418762 Chr.5: 69509190-69509190 on Build GRCh38
rs890382090 Chr.5: 69509181-69509181 on Build GRCh38
rs748899545 Chr.5: 69509103-69509103 on Build GRCh38
rs758735008 Chr.5: 69509198-69509198 on Build GRCh38
rs368281009 Chr.5: 69509146-69509146 on Build GRCh38
rs1044867212 Chr.5: 69509052-69509052 on Build GRCh38
rs201027704 Chr.5: 69509207-69509207 on Build GRCh38
rs761061240 Chr.5: 69509158-69509158 on Build GRCh38
rs888504593 Chr.5: 69509053-69509053 on Build GRCh38
rs147125035 Chr.5: 69509160-69509160 on Build GRCh38
rs202196960 Chr.5: 69509111-69509111 on Build GRCh38
rs1036844240 Chr.5: 69509057-69509057 on Build GRCh38
rs1029605974 Chr.5: 69509133-69509133 on Build GRCh38
rs200028558 Chr.5: 69509099-69509099 on Build GRCh38
rs941429315 Chr.5: 69509054-69509054 on Build GRCh38
rs768058632 Chr.5: 69509106-69509106 on Build GRCh38
rs759591993 Chr.5: 69509175-69509175 on Build GRCh38
rs898963712 Chr.5: 69509129-69509129 on Build GRCh38
rs371307537 Chr.5: 69509090-69509090 on Build GRCh38
rs374099764 Chr.5: 69509219-69509219 on Build GRCh38
rs373915080 Chr.5: 69509196-69509196 on Build GRCh38
rs750291439 Chr.5: 69509206-69509206 on Build GRCh38
rs370812139 Chr.5: 69509147-69509147 on Build GRCh38
rs776567486 Chr.5: 69509168-69509168 on Build GRCh38
rs201821077 Chr.5: 69509202-69509202 on Build GRCh38

View Full Product Details