Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41584518-41584743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PMM1
Entrez Gene ID
5372
Gene Name
phosphomannomutase 1
Gene Aliases
Sec53
UniGene
Hs.75835
Gene Chromosome Location
Chr.22: 41576886-41589867 on Build GRCh38
Transcripts
NM_002676.2 XM_005261638.4 XM_011530229.1 XM_011530230.2 XM_011530231.2 XM_011530232.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3554950 c.245C>T p.T82M Chr.22: 41584564-41584564 on Build GRCh38
6533306 c.273C>G p.L91L Chr.22: 41584536-41584536 on Build GRCh38
419528 c.261C>T p.H87H Chr.22: 41584548-41584548 on Build GRCh38
4941451 c.233C>T p.A78V Chr.22: 41584576-41584576 on Build GRCh38
6196113 c.234C>T p.A78A Chr.22: 41584575-41584575 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763520750 Chr.22: 41584547-41584547 on Build GRCh38
rs941482385 Chr.22: 41584688-41584688 on Build GRCh38
rs766611904 Chr.22: 41584548-41584548 on Build GRCh38
rs905953380 Chr.22: 41584672-41584672 on Build GRCh38
rs777030449 Chr.22: 41584542-41584542 on Build GRCh38
rs979158615 Chr.22: 41584577-41584577 on Build GRCh38
rs770624298 Chr.22: 41584646-41584646 on Build GRCh38
rs140192526 Chr.22: 41584569-41584569 on Build GRCh38
rs759675345 Chr.22: 41584634-41584634 on Build GRCh38
rs540695717 Chr.22: 41584545-41584545 on Build GRCh38
rs769928678 Chr.22: 41584650-41584650 on Build GRCh38
rs56017106 Chr.22: 41584645-41584645 on Build GRCh38
rs368220949 Chr.22: 41584636-41584636 on Build GRCh38
rs146168181 Chr.22: 41584563-41584563 on Build GRCh38
rs756930819 Chr.22: 41584593-41584593 on Build GRCh38
rs745415910 Chr.22: 41584665-41584665 on Build GRCh38
rs764141749 Chr.22: 41584636-41584637 on Build GRCh38
rs776917759 Chr.22: 41584602-41584603 on Build GRCh38
rs773372385 Chr.22: 41584651-41584651 on Build GRCh38
rs764459233 Chr.22: 41584564-41584564 on Build GRCh38
rs950041912 Chr.22: 41584598-41584598 on Build GRCh38
rs1056682478 Chr.22: 41584664-41584664 on Build GRCh38
rs199693358 Chr.22: 41584543-41584543 on Build GRCh38
rs533303728 Chr.22: 41584627-41584627 on Build GRCh38
rs992500885 Chr.22: 41584584-41584584 on Build GRCh38
rs778413259 Chr.22: 41584591-41584591 on Build GRCh38
rs142212781 Chr.22: 41584574-41584574 on Build GRCh38
rs750265458 Chr.22: 41584600-41584600 on Build GRCh38
rs779668310 Chr.22: 41584626-41584626 on Build GRCh38
rs376284083 Chr.22: 41584575-41584575 on Build GRCh38
rs770052672 Chr.22: 41584544-41584544 on Build GRCh38
rs758395507 Chr.22: 41584622-41584622 on Build GRCh38
rs939847296 Chr.22: 41584713-41584713 on Build GRCh38

View Full Product Details