Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10631846-10632077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH3
Entrez Gene ID
4621
Gene Name
myosin, heavy chain 3, skeletal muscle, embryonic
Gene Aliases
DA2A, DA2B, DA8, HEMHC, MYHC-EMB, MYHSE1, SMHCE
UniGene
Hs.440895
Gene Chromosome Location
Chr.17: 10628526-10679657 on Build GRCh38
Transcripts
NM_002470.3 XM_011523870.2 XM_011523871.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7315696 c.5062G>A p.E1688K Chr.17: 10631911-10631911 on Build GRCh38
5385663 c.4983C>T p.A1661A Chr.17: 10631990-10631990 on Build GRCh38
5954292 c.4962G>C p.T1654T Chr.17: 10632011-10632011 on Build GRCh38
1216211 c.5064G>C p.E1688D Chr.17: 10631909-10631909 on Build GRCh38
5385662 c.5079G>A p.L1693L Chr.17: 10631894-10631894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998354107 Chr.17: 10631881-10631881 on Build GRCh38
rs866198608 Chr.17: 10631924-10631924 on Build GRCh38
rs111358355 Chr.17: 10631885-10631885 on Build GRCh38
rs144450260 Chr.17: 10631984-10631984 on Build GRCh38
rs770715754 Chr.17: 10631902-10631902 on Build GRCh38
rs777159320 Chr.17: 10631986-10631986 on Build GRCh38
rs748302259 Chr.17: 10631960-10631960 on Build GRCh38
rs144771730 Chr.17: 10631868-10631868 on Build GRCh38
rs528933675 Chr.17: 10631911-10631911 on Build GRCh38
rs867393905 Chr.17: 10631913-10631913 on Build GRCh38
rs546075871 Chr.17: 10631865-10631865 on Build GRCh38
rs963650227 Chr.17: 10631948-10631948 on Build GRCh38
rs755136808 Chr.17: 10631875-10631875 on Build GRCh38
rs763805673 Chr.17: 10631920-10631920 on Build GRCh38
rs770789998 Chr.17: 10631985-10631985 on Build GRCh38
rs564468476 Chr.17: 10631886-10631886 on Build GRCh38
rs774366569 Chr.17: 10631904-10631904 on Build GRCh38
rs902245017 Chr.17: 10631872-10631872 on Build GRCh38
rs745783603 Chr.17: 10632002-10632002 on Build GRCh38
rs758929619 Chr.17: 10631905-10631905 on Build GRCh38
rs749111236 Chr.17: 10631900-10631900 on Build GRCh38
rs751923434 Chr.17: 10632035-10632035 on Build GRCh38
rs781405511 Chr.17: 10631955-10631955 on Build GRCh38
rs927954243 Chr.17: 10632015-10632015 on Build GRCh38
rs190188728 Chr.17: 10632012-10632012 on Build GRCh38
rs114770362 Chr.17: 10631864-10631864 on Build GRCh38
rs147973323 Chr.17: 10632011-10632011 on Build GRCh38
rs568057910 Chr.17: 10631910-10631910 on Build GRCh38
rs747006068 Chr.17: 10631874-10631874 on Build GRCh38
rs766750914 Chr.17: 10631943-10631943 on Build GRCh38
rs755587478 Chr.17: 10631954-10631954 on Build GRCh38
rs111742316 Chr.17: 10631927-10631927 on Build GRCh38
rs766741027 Chr.17: 10632033-10632033 on Build GRCh38
rs539678785 Chr.17: 10632057-10632057 on Build GRCh38
rs748986745 Chr.17: 10631982-10631982 on Build GRCh38
rs912678532 Chr.17: 10631980-10631980 on Build GRCh38
rs140180067 Chr.17: 10631899-10631899 on Build GRCh38
rs149260479 Chr.17: 10631921-10631921 on Build GRCh38
rs2239936 Chr.17: 10632032-10632032 on Build GRCh38
rs112858125 Chr.17: 10631906-10631906 on Build GRCh38
rs979326011 Chr.17: 10631934-10631934 on Build GRCh38
rs778068837 Chr.17: 10631966-10631966 on Build GRCh38
rs760113280 Chr.17: 10632044-10632044 on Build GRCh38
rs985838125 Chr.17: 10631978-10631978 on Build GRCh38

View Full Product Details