Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 130879537-130879808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX19
Entrez Gene ID
399979
Gene Name
sorting nexin 19
Gene Aliases
CHET8
UniGene
Hs.444024
Gene Chromosome Location
Chr.11: 130866254-130916488 on Build GRCh38
Transcripts
NM_001301089.1 NM_014758.2 XM_005271546.4 XM_011542819.2 XM_011542820.2 XM_011542821.2 XM_011542824.2 XM_011542825.2 XM_011542826.2 XM_011542827.2 XM_017017725.1 XM_017017726.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
925018 c.2828A>G p.Q943R Chr.11: 130879642-130879642 on Build GRCh38
925019 c.2776C>A p.L926I Chr.11: 130879694-130879694 on Build GRCh38
353735 c.2795G>T p.R932L Chr.11: 130879675-130879675 on Build GRCh38
2089554 c.2776C>T p.L926F Chr.11: 130879694-130879694 on Build GRCh38
4600053 c.2836C>G p.L946V Chr.11: 130879634-130879634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764361180 Chr.11: 130879669-130879669 on Build GRCh38
rs762043738 Chr.11: 130879752-130879752 on Build GRCh38
rs145177899 Chr.11: 130879745-130879745 on Build GRCh38
rs749648607 Chr.11: 130879753-130879753 on Build GRCh38
rs748843898 Chr.11: 130879720-130879720 on Build GRCh38
rs748354054 Chr.11: 130879634-130879634 on Build GRCh38
rs755044808 Chr.11: 130879609-130879610 on Build GRCh38
rs374167325 Chr.11: 130879692-130879692 on Build GRCh38
rs759295962 Chr.11: 130879665-130879665 on Build GRCh38
rs1030362205 Chr.11: 130879748-130879748 on Build GRCh38
rs753929758 Chr.11: 130879573-130879573 on Build GRCh38
rs771766375 Chr.11: 130879743-130879743 on Build GRCh38
rs1053067660 Chr.11: 130879731-130879731 on Build GRCh38
rs766043253 Chr.11: 130879658-130879658 on Build GRCh38
rs868354400 Chr.11: 130879664-130879664 on Build GRCh38
rs758703665 Chr.11: 130879594-130879594 on Build GRCh38
rs558566011 Chr.11: 130879609-130879609 on Build GRCh38
rs1008245013 Chr.11: 130879773-130879773 on Build GRCh38
rs776196813 Chr.11: 130879627-130879627 on Build GRCh38
rs373977709 Chr.11: 130879623-130879623 on Build GRCh38
rs772875754 Chr.11: 130879715-130879715 on Build GRCh38
rs948335200 Chr.11: 130879663-130879663 on Build GRCh38
rs368887462 Chr.11: 130879732-130879732 on Build GRCh38
rs763001221 Chr.11: 130879653-130879653 on Build GRCh38
rs773666603 Chr.11: 130879743-130879744 on Build GRCh38
rs765313539 Chr.11: 130879686-130879686 on Build GRCh38
rs746603028 Chr.11: 130879606-130879606 on Build GRCh38
rs747379345 Chr.11: 130879706-130879706 on Build GRCh38
rs746377779 Chr.11: 130879591-130879591 on Build GRCh38
rs199592034 Chr.11: 130879634-130879634 on Build GRCh38
rs757383273 Chr.11: 130879577-130879577 on Build GRCh38
rs774160457 Chr.11: 130879660-130879660 on Build GRCh38
rs769885066 Chr.11: 130879668-130879668 on Build GRCh38
rs758976076 Chr.11: 130879694-130879694 on Build GRCh38
rs773219751 Chr.11: 130879644-130879644 on Build GRCh38
rs561247244 Chr.11: 130879598-130879598 on Build GRCh38
rs774210019 Chr.11: 130879740-130879740 on Build GRCh38
rs780467524 Chr.11: 130879702-130879702 on Build GRCh38
rs373105417 Chr.11: 130879707-130879707 on Build GRCh38
rs566082292 Chr.11: 130879579-130879579 on Build GRCh38
rs769498026 Chr.11: 130879639-130879639 on Build GRCh38
rs765260422 Chr.11: 130879757-130879757 on Build GRCh38
rs200326824 Chr.11: 130879741-130879741 on Build GRCh38
rs368460883 Chr.11: 130879675-130879675 on Build GRCh38
rs780895525 Chr.11: 130879709-130879709 on Build GRCh38
rs371936742 Chr.11: 130879676-130879676 on Build GRCh38
rs1056096816 Chr.11: 130879620-130879620 on Build GRCh38
rs750471912 Chr.11: 130879761-130879761 on Build GRCh38
rs769375377 Chr.11: 130879713-130879713 on Build GRCh38
rs934482974 Chr.11: 130879716-130879716 on Build GRCh38
rs781302745 Chr.11: 130879621-130879622 on Build GRCh38

View Full Product Details