Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32683777-32684034 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDS5B
Entrez Gene ID
23047
Gene Name
PDS5 cohesin associated factor B
Gene Aliases
APRIN, AS3, CG008
UniGene
Hs.744901
Gene Chromosome Location
Chr.13: 32586427-32778020 on Build GRCh38
Transcripts
NM_015032.3 XM_005266298.4 XM_011534999.2 XM_011535000.2 XM_011535002.2 XM_017020448.1 XM_017020449.1 XM_017020450.1 XM_017020451.1 XM_017020452.1 XM_017020453.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5496236 c.1150C>G p.L384V Chr.13: 32683970-32683970 on Build GRCh38
946924 c.1202G>A p.R401Q Chr.13: 32684022-32684022 on Build GRCh38
5791133 c.1203+6T>C p.? Chr.13: 32684029-32684029 on Build GRCh38
5016425 c.1163_1166delACTT p.L390fs*3 Chr.13: 32683983-32683986 on Build GRCh38
946922 c.1086T>C p.P362P Chr.13: 32683906-32683906 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762151880 Chr.13: 32683977-32683977 on Build GRCh38
rs779507292 Chr.13: 32683869-32683869 on Build GRCh38
rs778463028 Chr.13: 32683889-32683889 on Build GRCh38
rs754611179 Chr.13: 32683882-32683882 on Build GRCh38
rs896631829 Chr.13: 32683936-32683936 on Build GRCh38
rs1004494165 Chr.13: 32683821-32683821 on Build GRCh38
rs768853340 Chr.13: 32683972-32683972 on Build GRCh38
rs201883179 Chr.13: 32683975-32683975 on Build GRCh38
rs777772241 Chr.13: 32683945-32683945 on Build GRCh38
rs375019534 Chr.13: 32683832-32683832 on Build GRCh38
rs758669346 Chr.13: 32683858-32683862 on Build GRCh38
rs542938836 Chr.13: 32683973-32683973 on Build GRCh38
rs372718271 Chr.13: 32683904-32683904 on Build GRCh38
rs771037519 Chr.13: 32683955-32683955 on Build GRCh38
rs750827034 Chr.13: 32684000-32684000 on Build GRCh38
rs767952800 Chr.13: 32683850-32683850 on Build GRCh38
rs1056663500 Chr.13: 32683933-32683933 on Build GRCh38
rs1012452699 Chr.13: 32683952-32683952 on Build GRCh38
rs1000960337 Chr.13: 32683984-32683984 on Build GRCh38
rs563682254 Chr.13: 32683833-32683833 on Build GRCh38
rs531281981 Chr.13: 32683964-32683964 on Build GRCh38
rs756679378 Chr.13: 32683860-32683860 on Build GRCh38
rs750915072 Chr.13: 32683856-32683856 on Build GRCh38
rs746943017 Chr.13: 32683947-32683947 on Build GRCh38
rs767862907 Chr.13: 32683983-32683983 on Build GRCh38
rs750550550 Chr.13: 32683830-32683831 on Build GRCh38
rs368394109 Chr.13: 32683897-32683897 on Build GRCh38
rs772008707 Chr.13: 32683927-32683927 on Build GRCh38
rs762971030 Chr.13: 32683971-32683971 on Build GRCh38

View Full Product Details