Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 50966205-50966462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPAG9
Entrez Gene ID
9043
Gene Name
sperm associated antigen 9
Gene Aliases
CT89, HLC-6, HLC4, HLC6, JIP-4, JIP4, JLP, PHET, PIG6
UniGene
Hs.463439
Gene Chromosome Location
Chr.17: 50962174-51120865 on Build GRCh38
Transcripts
NM_001130527.2 NM_001130528.2 NM_001251971.1 NM_003971.5 XM_005257768.2 XM_005257771.2 XM_005257774.3 XM_017025283.1 XM_017025284.1 XM_017025285.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1303049 c.3827C>T p.S1276L Chr.17: 50966369-50966369 on Build GRCh38
5819316 c.3885A>T p.K1295N Chr.17: 50966323-50966323 on Build GRCh38
4810611 c.3839C>T p.S1280L Chr.17: 50966369-50966369 on Build GRCh38
7391556 c.3886G>A p.D1296N Chr.17: 50966352-50966352 on Build GRCh38
4783662 c.3927C>T p.G1309G Chr.17: 50966281-50966281 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375541707 Chr.17: 50966358-50966358 on Build GRCh38
rs780201057 Chr.17: 50966379-50966379 on Build GRCh38
rs916244860 Chr.17: 50966267-50966267 on Build GRCh38
rs79122741 Chr.17: 50966432-50966432 on Build GRCh38
rs971996177 Chr.17: 50966288-50966288 on Build GRCh38
rs774780393 Chr.17: 50966287-50966287 on Build GRCh38
rs780408934 Chr.17: 50966400-50966402 on Build GRCh38
rs537149190 Chr.17: 50966249-50966249 on Build GRCh38
rs372214595 Chr.17: 50966353-50966353 on Build GRCh38
rs758634792 Chr.17: 50966364-50966364 on Build GRCh38
rs201214240 Chr.17: 50966425-50966425 on Build GRCh38
rs755129448 Chr.17: 50966391-50966391 on Build GRCh38
rs751540635 Chr.17: 50966390-50966390 on Build GRCh38
rs367786808 Chr.17: 50966308-50966308 on Build GRCh38
rs76182731 Chr.17: 50966426-50966426 on Build GRCh38
rs769936495 Chr.17: 50966417-50966417 on Build GRCh38
rs369871190 Chr.17: 50966409-50966409 on Build GRCh38
rs928911880 Chr.17: 50966302-50966302 on Build GRCh38
rs761394884 Chr.17: 50966349-50966349 on Build GRCh38
rs568928274 Chr.17: 50966242-50966242 on Build GRCh38
rs371771793 Chr.17: 50966239-50966239 on Build GRCh38
rs367859497 Chr.17: 50966235-50966235 on Build GRCh38
rs774903112 Chr.17: 50966434-50966434 on Build GRCh38
rs958798752 Chr.17: 50966256-50966256 on Build GRCh38
rs374188076 Chr.17: 50966240-50966240 on Build GRCh38
rs760234541 Chr.17: 50966293-50966293 on Build GRCh38
rs9896965 Chr.17: 50966279-50966279 on Build GRCh38
rs758211942 Chr.17: 50966366-50966366 on Build GRCh38
rs753566343 Chr.17: 50966336-50966336 on Build GRCh38
rs148354979 Chr.17: 50966281-50966281 on Build GRCh38
rs781113712 Chr.17: 50966401-50966401 on Build GRCh38

View Full Product Details