Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 127599411-127599654 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCM2
Entrez Gene ID
4171
Gene Name
minichromosome maintenance complex component 2
Gene Aliases
BM28, CCNL1, CDCL1, D3S3194, DFNA70, MITOTIN, cdc19
UniGene
Hs.477481
Gene Chromosome Location
Chr.3: 127598357-127622436 on Build GRCh38
Transcripts
NM_004526.3 NR_073375.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7223850 c.173G>T p.G58V Chr.3: 127599484-127599484 on Build GRCh38
3059183 c.116C>T p.T39I Chr.3: 127599427-127599427 on Build GRCh38
6709779 c.101G>A p.R34H Chr.3: 127599412-127599412 on Build GRCh38
6317911 c.150G>T p.E50D Chr.3: 127599461-127599461 on Build GRCh38
1037909 c.141A>G p.P47P Chr.3: 127599452-127599452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780508768 Chr.3: 127599458-127599463 on Build GRCh38
rs551454910 Chr.3: 127599581-127599581 on Build GRCh38
rs765659379 Chr.3: 127599479-127599479 on Build GRCh38
rs749119167 Chr.3: 127599542-127599542 on Build GRCh38
rs757457211 Chr.3: 127599509-127599509 on Build GRCh38
rs796576308 Chr.3: 127599524-127599524 on Build GRCh38
rs771946607 Chr.3: 127599471-127599471 on Build GRCh38
rs774376718 Chr.3: 127599570-127599570 on Build GRCh38
rs985682013 Chr.3: 127599615-127599615 on Build GRCh38
rs768595192 Chr.3: 127599609-127599609 on Build GRCh38
rs575839692 Chr.3: 127599594-127599594 on Build GRCh38
rs772694646 Chr.3: 127599590-127599590 on Build GRCh38
rs3087452 Chr.3: 127599515-127599515 on Build GRCh38
rs770134211 Chr.3: 127599539-127599539 on Build GRCh38
rs768283316 Chr.3: 127599566-127599566 on Build GRCh38
rs1001619429 Chr.3: 127599586-127599586 on Build GRCh38
rs374462347 Chr.3: 127599597-127599597 on Build GRCh38
rs761404358 Chr.3: 127599455-127599455 on Build GRCh38
rs746153512 Chr.3: 127599528-127599528 on Build GRCh38
rs751974456 Chr.3: 127599507-127599507 on Build GRCh38
rs758633607 Chr.3: 127599491-127599491 on Build GRCh38
rs778367347 Chr.3: 127599493-127599493 on Build GRCh38
rs957445179 Chr.3: 127599599-127599599 on Build GRCh38
rs376685268 Chr.3: 127599598-127599598 on Build GRCh38
rs375851208 Chr.3: 127599441-127599441 on Build GRCh38
rs572319263 Chr.3: 127599470-127599470 on Build GRCh38
rs140680894 Chr.3: 127599437-127599437 on Build GRCh38
rs780354621 Chr.3: 127599540-127599540 on Build GRCh38
rs760296226 Chr.3: 127599477-127599477 on Build GRCh38
rs775414015 Chr.3: 127599440-127599440 on Build GRCh38
rs771618507 Chr.3: 127599589-127599589 on Build GRCh38
rs572334760 Chr.3: 127599442-127599442 on Build GRCh38
rs201221422 Chr.3: 127599511-127599511 on Build GRCh38
rs775184023 Chr.3: 127599513-127599513 on Build GRCh38
rs753111466 Chr.3: 127599480-127599480 on Build GRCh38

View Full Product Details