Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 55343773-55344010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR4A16
Entrez Gene ID
81327
Gene Name
olfactory receptor family 4 subfamily A member 16
Gene Aliases
OR11-117, OR4A16Q
UniGene
Hs.554530
Gene Chromosome Location
Chr.11: 55343201-55344187 on Build GRCh38
Transcripts
NM_001005274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6734345 c.789T>C p.F263F Chr.11: 55343989-55343989 on Build GRCh38
7246276 c.688G>T p.E230* Chr.11: 55343888-55343888 on Build GRCh38
3967577 c.740T>C p.L247P Chr.11: 55343940-55343940 on Build GRCh38
4506420 c.719C>T p.S240F Chr.11: 55343919-55343919 on Build GRCh38
4572108 c.646T>G p.C216G Chr.11: 55343846-55343846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748490235 Chr.11: 55343814-55343814 on Build GRCh38
rs79231280 Chr.11: 55343977-55343977 on Build GRCh38
rs535471161 Chr.11: 55343913-55343913 on Build GRCh38
rs767128231 Chr.11: 55343910-55343910 on Build GRCh38
rs199539632 Chr.11: 55343949-55343949 on Build GRCh38
rs374940663 Chr.11: 55343952-55343952 on Build GRCh38
rs145729272 Chr.11: 55343813-55343813 on Build GRCh38
rs556673264 Chr.11: 55343902-55343902 on Build GRCh38
rs201025638 Chr.11: 55343880-55343880 on Build GRCh38
rs755901185 Chr.11: 55343978-55343978 on Build GRCh38
rs12807769 Chr.11: 55343895-55343895 on Build GRCh38
rs373240242 Chr.11: 55343840-55343840 on Build GRCh38
rs772252213 Chr.11: 55343830-55343830 on Build GRCh38
rs267602962 Chr.11: 55343854-55343854 on Build GRCh38
rs78131590 Chr.11: 55343929-55343929 on Build GRCh38
rs747967582 Chr.11: 55343931-55343931 on Build GRCh38
rs773223799 Chr.11: 55343933-55343933 on Build GRCh38
rs756483302 Chr.11: 55343874-55343874 on Build GRCh38
rs79376849 Chr.11: 55343822-55343822 on Build GRCh38
rs145541491 Chr.11: 55343849-55343849 on Build GRCh38
rs750752095 Chr.11: 55343836-55343836 on Build GRCh38
rs749923481 Chr.11: 55343852-55343852 on Build GRCh38
rs369347524 Chr.11: 55343839-55343839 on Build GRCh38
rs761558050 Chr.11: 55343897-55343897 on Build GRCh38
rs200720954 Chr.11: 55343837-55343837 on Build GRCh38
rs766122369 Chr.11: 55343865-55343865 on Build GRCh38
rs764807240 Chr.11: 55343806-55343806 on Build GRCh38
rs745945050 Chr.11: 55343885-55343885 on Build GRCh38
rs966230142 Chr.11: 55343927-55343927 on Build GRCh38
rs78534550 Chr.11: 55343817-55343817 on Build GRCh38
rs779463185 Chr.11: 55343914-55343914 on Build GRCh38
rs761841567 Chr.11: 55343842-55343842 on Build GRCh38
rs376009802 Chr.11: 55343946-55343946 on Build GRCh38
rs548070218 Chr.11: 55343858-55343858 on Build GRCh38
rs141072345 Chr.11: 55343801-55343801 on Build GRCh38
rs769532333 Chr.11: 55343932-55343932 on Build GRCh38
rs75741638 Chr.11: 55343889-55343889 on Build GRCh38
rs745983959 Chr.11: 55343823-55343823 on Build GRCh38
rs756663590 Chr.11: 55343799-55343799 on Build GRCh38
rs267602963 Chr.11: 55343920-55343920 on Build GRCh38
rs758018143 Chr.11: 55343857-55343857 on Build GRCh38
rs750886536 Chr.11: 55343953-55343953 on Build GRCh38
rs770462778 Chr.11: 55343848-55343848 on Build GRCh38
rs780189900 Chr.11: 55343890-55343890 on Build GRCh38
rs776507750 Chr.11: 55343896-55343896 on Build GRCh38
rs75076309 Chr.11: 55343878-55343878 on Build GRCh38
rs200486306 Chr.11: 55343903-55343903 on Build GRCh38
rs74668728 Chr.11: 55343937-55343937 on Build GRCh38
rs78913302 Chr.11: 55343906-55343906 on Build GRCh38
rs780494375 Chr.11: 55343882-55343882 on Build GRCh38
rs758924632 Chr.11: 55343955-55343955 on Build GRCh38
rs751607084 Chr.11: 55343957-55343957 on Build GRCh38
rs749809790 Chr.11: 55343821-55343821 on Build GRCh38
rs76998459 Chr.11: 55343930-55343930 on Build GRCh38
rs189929394 Chr.11: 55343962-55343962 on Build GRCh38
rs779330023 Chr.11: 55343883-55343883 on Build GRCh38
rs770520415 Chr.11: 55343934-55343934 on Build GRCh38
rs780583330 Chr.11: 55343956-55343956 on Build GRCh38
rs781461278 Chr.11: 55343973-55343973 on Build GRCh38
rs747520222 Chr.11: 55343893-55343893 on Build GRCh38
rs778086947 Chr.11: 55343820-55343820 on Build GRCh38
rs758142642 Chr.11: 55343809-55343809 on Build GRCh38
rs141537673 Chr.11: 55343942-55343942 on Build GRCh38
rs758404250 Chr.11: 55343928-55343928 on Build GRCh38
rs149774017 Chr.11: 55343808-55343808 on Build GRCh38
rs368388243 Chr.11: 55343900-55343900 on Build GRCh38
rs751311802 Chr.11: 55343867-55343867 on Build GRCh38

View Full Product Details