Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69419441-69419712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NFU1
Entrez Gene ID
27247
Gene Name
NFU1 iron-sulfur cluster scaffold
Gene Aliases
CGI-33, HIRIP, HIRIP5, MMDS1, NIFUC, Nfu, NifU
UniGene
Hs.430439
Gene Chromosome Location
Chr.2: 69396113-69438123 on Build GRCh38
Transcripts
NM_001002755.2 NM_001002756.2 NM_015700.3 NR_045631.1 NR_045632.1 XM_017003808.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5938913 c.369+1G>A p.? Chr.2: 69419537-69419537 on Build GRCh38
1022415 c.339C>A p.F113L Chr.2: 69419568-69419568 on Build GRCh38
276331 c.354C>A p.F118L Chr.2: 69419553-69419553 on Build GRCh38
5823685 c.348A>T p.P116P Chr.2: 69419559-69419559 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775249424 Chr.2: 69419534-69419534 on Build GRCh38
rs748373355 Chr.2: 69419501-69419501 on Build GRCh38
rs778091932 Chr.2: 69419614-69419614 on Build GRCh38
rs760301873 Chr.2: 69419650-69419650 on Build GRCh38
rs1032794033 Chr.2: 69419476-69419476 on Build GRCh38
rs779031661 Chr.2: 69419496-69419496 on Build GRCh38
rs371546359 Chr.2: 69419606-69419606 on Build GRCh38
rs756434076 Chr.2: 69419575-69419575 on Build GRCh38
rs200341023 Chr.2: 69419615-69419615 on Build GRCh38
rs896494878 Chr.2: 69419685-69419685 on Build GRCh38
rs574127236 Chr.2: 69419678-69419678 on Build GRCh38
rs748188386 Chr.2: 69419604-69419604 on Build GRCh38
rs753851157 Chr.2: 69419608-69419609 on Build GRCh38
rs955258748 Chr.2: 69419528-69419528 on Build GRCh38
rs772895386 Chr.2: 69419648-69419648 on Build GRCh38
rs369585960 Chr.2: 69419522-69419522 on Build GRCh38
rs763572439 Chr.2: 69419538-69419538 on Build GRCh38
rs755494964 Chr.2: 69419598-69419598 on Build GRCh38
rs761264823 Chr.2: 69419639-69419639 on Build GRCh38
rs778924871 Chr.2: 69419602-69419602 on Build GRCh38
rs757661442 Chr.2: 69419559-69419559 on Build GRCh38
rs372898848 Chr.2: 69419616-69419617 on Build GRCh38
rs767100000 Chr.2: 69419560-69419560 on Build GRCh38
rs749252242 Chr.2: 69419628-69419628 on Build GRCh38
rs750812769 Chr.2: 69419573-69419573 on Build GRCh38
rs764082701 Chr.2: 69419526-69419526 on Build GRCh38
rs1001185238 Chr.2: 69419608-69419608 on Build GRCh38
rs552142580 Chr.2: 69419552-69419552 on Build GRCh38
rs768661373 Chr.2: 69419487-69419487 on Build GRCh38
rs767192006 Chr.2: 69419640-69419640 on Build GRCh38
rs768573293 Chr.2: 69419630-69419630 on Build GRCh38
rs769873909 Chr.2: 69419533-69419533 on Build GRCh38
rs773747897 Chr.2: 69419637-69419637 on Build GRCh38
rs755149278 Chr.2: 69419496-69419498 on Build GRCh38
rs1053163969 Chr.2: 69419488-69419488 on Build GRCh38
rs751077770 Chr.2: 69419550-69419550 on Build GRCh38
rs538875159 Chr.2: 69419504-69419504 on Build GRCh38
rs769749096 Chr.2: 69419622-69419622 on Build GRCh38
rs776568167 Chr.2: 69419520-69419520 on Build GRCh38
rs569145992 Chr.2: 69419594-69419594 on Build GRCh38
rs964033662 Chr.2: 69419509-69419509 on Build GRCh38
rs1021842189 Chr.2: 69419579-69419579 on Build GRCh38
rs141033711 Chr.2: 69419623-69419623 on Build GRCh38

View Full Product Details